Skip to Content
Merck
All Photos(1)

Key Documents

ST1551

Sigma-Aldrich

Anti-GGT1 Mouse mAb (1F9)

liquid, clone 1F9, Calbiochem®

Synonym(s):

Anti-γ-Glutamyltransferase1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$1,080.00

$1,080.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$1,080.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$1,080.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F9, monoclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2α

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GGT1(2678)

General description

Anti-GGT1, mouse monoclonal, clone 1F9, recognizes the ~60 kDa GGT1 protein in NIH3T3 cells and human testes tissue. It is validated for Western blotting & immunohistochemistry on paraffin sections.
Purified mouse monoclonal antibody. Recognizes the ~60 kDa GGT1 protein.
Recognizes the ~60 kDa GGT1 protein in NIH3T3 cells and human testes tissue.

Immunogen

A recombinant polypeptide corresponding to amino acids 381-471 of human GGT1, expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Analysis Note

Positive Control
NIH3T3 cells, Human testes tissue

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julieta N Caballero et al.
PloS one, 8(6), e65364-e65364 (2013-06-21)
Acquisition of fertilization ability by spermatozoa during epididymal transit occurs in part by the transfer of molecules from membranous vesicles called epididymosomes. Epididymosomes are heterogeneous in terms of both size and molecular composition. Exosomes and other related small membranous vesicles
Shenaz Khan et al.
JCI insight, 5(15) (2020-07-03)
Kidney disease is one of the most devastating complications of diabetes, and tubular atrophy predicts diabetic kidney disease (DKD) progression to end-stage renal disease. We have proposed that fatty acids bound to albumin contribute to tubular atrophy by inducing lipotoxicity

Questions

  1. Can we use this antibody on bovine y-spermatozoa to bind surface protein of sperm cell ?

    1 answer
    1. The immunogen of ST1551 Sigma-Aldrich Anti-GGT1 Mouse mAb (1F9) is a recombinant polypeptide corresponding to amino acids 381-471 of human GGT1, expressed as a GST fusion protein. This sequence has 89% homology to bovine, so most likely the antibody will work in bovine, but we have not tested it in-house in bovine samples.

      It appears that ST1551 has been utilized with bovine spermatozoa in the papers referenced below. It is recommended to review these papers for detailed information regarding the experimental protocols and results.
      CD9-Positive Microvesicles Mediate the Transfer of Molecules to Bovine Spermatozoa during Epididymal Maturation
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC3681974/

      https://www.google.com/search?q=ST1551+bovine&rlz=1C1GCEA_enUS976US976&oq=ST1551+bovine&gs_lcrp=EgZjaHJvbWUyBggAEEUYOTIGCAEQRRg90gEIMzE5OWowajeoAgCwAgA&sourceid=chrome&ie=UTF-8#ip=1

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service