Skip to Content
Merck
All Photos(1)

Key Documents

ST1049

Sigma-Aldrich

Anti-20S Proteasome α1, 2, 3, 5, 6, & 7-Subunits Mouse mAb (MCP231)

liquid, clone MCP231, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MCP231, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, yeast, rat, rabbit, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSMA1(5682)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with mouse myleloma cells. Recognizes the ~29-32 kDa α1, 2, 3, 5, 6 and 7 subunits of the 20S proteasome.
Recognizes α1, 2, 3, 5, 6 and 7 subunits of 20S proteasome in human placental proteasome preparations.
This Anti-20S Proteasome alpha-1, 2, 3, 5, 6, & 7 Subunits Antibody is validated for use in WB, IP, ELISA, IF, ICC and IHC for the detection of 20S Proteasome alpha-1, 2, 3, 5, 6, & 7 Subunits.

Immunogen

Epitope: within the prosbox I motif common to α subunits (TVWSPQGRLHQVEYAMEA)
dinitrophenylated proteasomes

Application


Immunoblotting (1:1000, chemiluminescence)
Immunoprecipitation (not recommended)
ELISA (see comments)
Immunofluorescence (see application references)
Immunocytochemistry (see application references)
Immunohistochemistry (see application references)
Frozen Sections (see application references)

Warning

Toxicity: Highly Toxic (H)

Analysis Note

Positive Control
Human placental tissue

Other Notes

Immunoblotting using a human placental proteasome preparation yields two bands at ~29 kDa and ~32 kDa. In ELISA the antibody reacts with the peptide sequence TVWSPQGRLHQVEYAMEA encompassing the prosbox I motif common to α-type subunits. Antibody should be titrated for optimal results in individual systems.
Kopp, F., et al. 1997. Proc. Natl. Acad. Sci. USA94, 2939.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine S Yanagi et al.
PLoS biology, 22(7), e3002720-e3002720 (2024-07-11)
The conserved SKN-1A/Nrf1 transcription factor regulates the expression of proteasome subunit genes and is essential for maintenance of adequate proteasome function in animal development, aging, and stress responses. Unusual among transcription factors, SKN-1A/Nrf1 is a glycoprotein synthesized in the endoplasmic
Lin Lyu et al.
Autophagy, 1-16 (2021-08-13)
Until recently, the ubiquitin-proteasome system (UPS) and macroautophagy/autophagy were considered to be two independent systems that target proteins for degradation by proteasomes or via lysosomes, respectively. Here, we report that TRIM44 (tripartite motif containing 44) is a novel link that
Shuangwu Sun et al.
Open biology, 7(5) (2017-05-26)
Ubiquitin-like domain-containing C-terminal domain phosphatase 1 (UBLCP1), an FCP/SCP phosphatase family member, was identified as the first proteasome phosphatase. UBLCP1 binds to proteasome subunit Rpn1 and dephosphorylates the proteasome in vitro However, it is still unclear which proteasome subunit(s) are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service