Skip to Content
Merck
All Photos(1)

Key Documents

SNAP2FRIHC

Millipore

SNAP i.d. 2.0 IHC Frame

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$1,900.00

$1,900.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$1,900.00

About This Item

UNSPSC Code:
41116010
eCl@ss:
42029053
NACRES:
NB.22

$1,900.00


Check Cart for Availability

manufacturer/tradename

SNAP id®

Quality Level

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunohistochemistry (frozen sections): suitable

compatibility

for use with Commercially available blocking reagents
for use with Luminata Western HRP Substrates
for use with Nitrocellulose
for use with PVDF (Immobilon membranes)
for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01)
for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01)
for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001)
for use with commercially available detection reagents

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

storage temp.

room temp

General description

The SNAP id® 2.0 System for IHC introduces a streamlined, medium through-put IHC capability to our SNAP i.d. 2.0 family. The frame and slide holders enable you to process up to 12 tissue slides, while our unique vacuum technology evenly removes your solutions - no more dipping or pouring.

Application

For use with immunohistochemistry slides.

Legal Information

SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

4.1B - Flammable solid hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service