Skip to Content
Merck
All Photos(1)

Key Documents

SCT277

Sigma-Aldrich

BioTracker Red pH-Independent Autophagy Live Cell Probe

Human

Synonym(s):

autophagosome live cell probe, red AP live cell probe, red autophagosome probe, red autophagy probe

Sign Into View Organizational & Contract Pricing

Select a Size

5 NMOL
$668.00

$668.00


Estimated to ship on28 May 2025



Select a Size

Change View
5 NMOL
$668.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.47

$668.00


Estimated to ship on28 May 2025


Product Name

BioTracker Red pH-Independent Autophagy Live Cell Probe,

biological source

human

Quality Level

packaging

vial of 5 nmol

manufacturer/tradename

Millipore

technique(s)

cell culture | stem cell: suitable
flow cytometry: suitable

shipped in

2-8°C

storage temp.

2-8°C

General description

The small fluorescent molecule SCT277 is used to detect autophagosomes and autolysosomes. The presumed mechanism is that the dye is incorporated into the autophagosome during double-membrane formation via structural features, and then emits fluorescence under hydrophobic conditions. The probe emits in the far red range.SPECTRAL CHARACTERISTICS: Excitation λ: 530 nmEmission λ: 720 nm

Features and Benefits

Fluorescence microscopyFlow cytometry

Target description

BioTracker Red pH-Independent Autophagy Live Cell Probe detects autophagosomes and autolysosomes. The presumptive mechanism involves incorporation of the probe via structural features into the autophagosome during double-membrane formation. Upon incorporation, the probe emits fluorescence under hydrophobic conditions. The utility of the probe is conferred by molecular properties that include cell permeability, does not require transfection, and enables live cell detection with fluorescence microscopy or other fluorescence capture instruments.

Storage and Stability

Store at -20 °C, protected from light.

Other Notes

SCT277 detects autophagosomes and autolysosomes by the likely mechanism of incorporation into the autophagosome during double membrane formation, and emits red fluorescence under hydrophobic conditions. SCT277 is cell permeable, does not require transfection, and enables live cell imaging with fluorescence microscopy and quantitative assay by flow cytometry.Add 50 uL DMSO to tube of SCT277 (5 nmol) and pipet to dissolve to make a 0.1 mM stock solution. Dilute with culture medium to 0.1 - 0.5 uM to make working solution.

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service