Skip to Content
Merck
All Photos(1)

Key Documents

SCT121

Millipore

GelRed® Nucleic Acid Stain 3X Water

GelRed is a fluorescent nucleic acid stain designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels.

Synonym(s):

Fluorescent DNA gel stain

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
$553.00

$553.00


Estimated to ship on25 May 2025



Select a Size

Change View
4 L
$553.00

About This Item

UNSPSC Code:
12352106
NACRES:
NA.52

$553.00


Estimated to ship on25 May 2025


form

liquid

technique(s)

PCR: suitable

solubility

water: soluble

detection method

fluorometric

General description

GelRed is a sensitive, stable and environmentally safe fluorescent nucleic acid dye designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels. GelRed and EtBr have virtually the same spectra (Figure 1), so you can directly replace EtBr with GelRed without changing your existing imaging system. In addition, GelRed is far more sensitive than EtBr (Figure 2).

The dye is noncytotoxic and non-mutagenic at concentrations well above the working concentrations used in gel staining. GelRed successfully passed environmental safety tests in compliance with CCR Title 22 Hazardous Waste Characterization, under which GelRed is not classified as hazardous waste.

Spectral Properties
Absorbance: Standard Transilluminator (302 or 312 nm)
Emission: Ethidium Bromide, SYBR or GelStar Filter

Application

GelRed is a fluorescent nucleic acid stain designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels.
Nucleic acid detection
Research Category
Cell Culture
Research Sub Category
Live Cell Dye

Quality

Spectral Properties
Absorbance: Standard Transilluminator (302 or 312 nm)
Emission: Ethidium Bromide, SYBR or GelStar Filter

Physical form

Liquid

Storage and Stability

GelRed is a very stable dye. Store GelRed at room temperature, protected from light. Dye precipitation may occur at lower temperatures, resulting in lower signal or the appearance of precipitate on the surface of the gel. If this occurs, heat the solution to 45-50°C for two minutes and vortex. Protect From Light.

Legal Information

GelRed is a registered trademark of Biotium Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service