Skip to Content
Merck
All Photos(1)

Key Documents

SCT102

Sigma-Aldrich

BioTracker NucView® 405 Blue Caspase-3 Dye (DMSO)

Live cell imaging apoptosis dye for caspase-3/7 enzyme activity used to detect programmed cell death using fluorescence microscopy or flow cytometry applications.

Synonym(s):

Caspase-3/7 apoptosis dye, Live cell imaging probe

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$824.00

$824.00


Estimated to ship on01 June 2025

Available for sale now


Select a Size

Change View
100 μL
$824.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

$824.00


Estimated to ship on01 June 2025

Available for sale now

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Caspases are a family of protease enzymes that play essential roles in programmed cell death, termed apoptosis. Caspase-3 is a caspase protein that interacts with caspase-8 and caspase-9 and sequential activation of plays a central role in the execution-phase of cellular apoptosis.

The BioTracker NucView 405 Blue Caspase-3 Dye provides a convenient live cell tool for profiling apoptotic cell populations based on caspase-3/7 activity using either fluorescence microscopy or flow cytometry. In contrast to other fluorogenic caspase substrates or fluorescent caspase inhibitor based (FLICA) assays, the BioTracker NucView 405 Caspase-3 Dye can be used to detect caspase-3/7 activity within individual intact cells without inhibiting apoptosis progression. The substrate consists of a fluorogenic DNA dye coupled to the caspase-3/7 DEVD recognition sequence. The substrate, which is initially non-fluorescent, penetrates the plasma membrane and enters the cytoplasm. In apoptotic cells, caspase-3/7 cleaves the substrate, releasing the high-affinity DNA dye, which migrates to the cell nucleus and stains DNA with bright blue fluorescence. Thus, NucView 405 Caspase-3 Substrate is bifunctional, allowing detection of caspase-3/7 activity and visualization of morphological changes in the nucleus during apoptosis. The dye is formaldehyde-fixable and compatible with subsequent immunostaining. The dye is offered in DMSO and PBS (phosphate-buffered saline) formulations. The substrate in PBS is formulated for use in cells that are sensitive to DMSO toxicity.

NucView 405 dye is excited by the 405nm laser line for detection by confocal microscopy in the DAPI channel, or by flow cytometry in the Pacific Blue channel. NucView 405 is ideal for caspase-3 detection in multi-color applications for researchers who wish to reserve the green fluorescence channel for other detection reagents.

Spectral Properties
Absorbance: 429nm
Emission: 469nm

Application

Live cell fluorescent imaging
Live cell imaging apoptosis dye for caspase-3/7 enzyme activity used to detect programmed cell death using fluorescence microscopy or flow cytometry applications.
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Quality

Spectral Properties
Absorbance: 429nm
Emission: 469nm

Physical form

Liquid

Storage and Stability

Store BioTracker NucView 405 Blue Caspase-3 Dye, 1mM in DMSO at 2-8ºC. Protect From Light.
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Legal Information

NucView is a registered trademark of Biotium Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can this dye be used for in introvital imaging of mice? What is the appropriate concentration? How long can I monitor the dye under the microscope? Is there any relevant work report?

    1 answer
    1. Unfortunately, this product has not been tested for use in intravital microscopy. A search of the literature did not reveal any related publications. The end user would have to determine suitability.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service