Skip to Content
Merck
All Photos(1)

Key Documents

SCT090

Sigma-Aldrich

BioTracker Bacterial FtsZ Live Cell Dye

Synonym(s):

BioTracker Bacterial FtsZ Live Cell Dye

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.47
Pricing and availability is not currently available.

form

lyophilized

mol wt

calculated mol wt 782.1

packaging

pkg of 1 mg

manufacturer/tradename

Millipore

storage condition

protect from light

growth mode

N/A

shipped in

ambient

storage temp.

−20°C

General description

The filamentous temperature-sensitive Z (FtsZ) protein is a promising target for the development of new antibacterial drug targets. FtsZ is a protein encoded by the ftsZ gene that assembles into a ring at the future site of bacterial cell division and is one of the most conserved proteins in prokaryotes. This live cell probe will aid development of new FtsZ inhibitors and enable screening for FtsZ inhibition in a broad range of bacterial species. BioTracker Bacterial FtsZ Live Cell Dye is a BODIPY conjugated fluorescent probe that binds FtsZ proteins from both Gram-positive pathogens (Staphylococcus aureus, Enterococcus faecalis, Enterococcus faecium, Streptococcus pyogenes, Streptococcus agalactiae, and Streptococcus pneumoniae) and Gram-negative pathogens (Escherichia coli, Klebsiella pneumoniae, Pseudomonas aeruginosa, and Acinetobacter baumannii) (1). It allows for visualization of FtsZ using fluorescence microscopy, and can be used to monitor and identify non-fluorescent FtsZ inhibitors in these target pathogens. Spectral PropertiesExcitation: 488 nmEmission: 510 nmReference:1. Ferrer-González, Edgar, et al. "Structure-Guided Design of a fluorescent probe for the Visualization of ftsZ in clinically important Gram-positive and Gram-negative Bacterial pathogens." Scientific reports 9.1 (2019): 1-16.

Application

Purity ≥ 98% confirmed by HPLC. Identification confirmed by HNMR, LC-MS, and elemental analysis.

Features and Benefits

Fluorescence probe for detecting FtsZ in bacteria

Physical form

Lyophilized. Red solid.

Storage and Stability

Store at -20°C, desiccate and protect from light.
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service