Skip to Content
Merck
All Photos(1)

Key Documents

SCT024

Sigma-Aldrich

BioTracker 543 Yellow β-Gal Dye

Live cell imaging dye for β-galactosidase (β-gal) used to detect LacZ reporter gene expression and cellular senescence (SA-β-gal).

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing

Select a Size

10 X 50 μG
$1,130.00

$1,130.00


Estimated to ship on31 May 2025



Select a Size

Change View
10 X 50 μG
$1,130.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.47

$1,130.00


Estimated to ship on31 May 2025


technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

β-galactosidase, also called beta-gal or β-gal, is a glycoside hydrolase enzyme that catalyzes the hydrolysis of β-galactosides into monosaccharides through the breaking of a glycosidic bond. β-gal is commonly used in molecular biology as a reporter marker to monitor gene expression using the chromogenic X-Gal. β-gal has also been used to measure cellular senescence (SA-β-gal).

The BioTracker 543 Yellow β-Gal Dye is a fluorescent probe for the detection of β-galactosidase in living cells. It can be applied to fluorescent imaging and selection of cell and tissue transfected with lacZ along as a measurement of cellular senescence. Other applications include gene analysis by fluorescent imaging, monitoring transfection efficiency, and the study of gene promoter or enhancer elements. Because the dye is almost non-fluorescent in the absence of β-galactosidase, this probe exhibits good S/N ratio.

Spectral Properties
Absorbance: 525nm (488nm compatible)
Emission: 543nm

Storage and Stability

Store BioTracker 543 Yellow β-Gal Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service