Skip to Content
Merck
All Photos(2)

Documents

SCR721

Sigma-Aldrich

TagRFP Simplicon® RNA (E3L) Kit

To determine optimal transfection conditions to express the self-replicating RNA of your interest through using Simplicon Cloning Vector (E3L) (Part #:SCR724) in hard-to- transfect somatic or primary cells

Synonym(s):

Self-replicating TagRFP RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.81

technique(s)

nucleic acid detection: suitable (RNA)

shipped in

ambient

General description

Simplicon is a novel system to effect immediate high sustained protein expression of multiple genes into transfected cells without the risk of genome integration. The technology employs a single, synthetic, polycistronic, self-replicating RNA based on the Venezuelan equine encephalitis (VEE) genome1,2,3. The Simplicon RNA contains only genes encoding the VEE RNA replication machinery while the structural proteins that are required to make an infectious particle have been removed and replaced with the transgenes of interest.

Introduction and replication of the Simplicon RNA is expected to elicit a strong interferon response in transfected cells. To suppress the IFN responses, a Vaccinia virus protein4, B18R, is used for the original Simplicon technology. Recently, we found that another Vaccinia virus protein4, E3L, also suppresses the IFN responses in Simplicon RNA expression. B18R neutralizes type I interferons by direct binding, while E3L inhibits the cytoplasmic signaling pathwaycs of IFN responses. Therefore, B18R and E3L are both employed in the Simplicon Expression System and work collaboratively to suppress IFN responses. As a result, there is increased cell viability during RNA transfection and increased expression of the transgenes. The Simplicon Expression System works in human cells and is not expected to work in mouse cells. This is because the B18R does not effectively neutralize mouse interferon (IFN)-β.

The TagRFP Simplicon RNA (E3L) Kit was developed to enable the evaluation of the Simplicon Expression System in targeted cells. E3L4 incorporation in the Simplicon Expression System strongly suppresses interferon responses and enables increased expression of the target gene(s). TagRFP Simplicon RNA expresses a monomeric red (orange) fluorescent protein generated from the RFP of sea anemone Entacmaea quadricolor. TagRFP exhibits fluorescence with excitation/ emission maxima at 555/584 nm respectively, and brightness that is nearly three times higher than mCherry5.6.

Application

The Simplicon TagRFP RNA (E3L) Kit was developed to enable the evaluation of the Simplicon expression system in your cell line(s). For example, it may be used to determine optimal transfection conditions to express the self-replicating RNA of your interest through using Simplicon Cloning Vector (E3L) (Part #:SCR724) in hard-to- transfect somatic or primary cells.

Components

1.TagRFP Simplicon RNA (E3L): (Cat. # CS224550) One (1) vial containing 10 µL of RNA (1 µg/µL). Store at -80°C.

2.B18R-E3L RNA: (Cat. # CS224503) One (1) vial containing 10 µL of RNA (1 µg/µL). Store at -80°C.

Quality

•Discrete RNA band on RNA gel of appropriate size.
•40-60% RFP-positive human foreskin fibroblasts (HFFs) with FACS analysis.

Storage and Stability

1.TagRFP SimpliconTM RNA (E3L): (Cat. # CS224550) One (1) vial containing 10 µL of RNA (1 µg/µL). Store at -80°C.

2.B18R-E3L RNA: (Cat. #CS224503) One (1) vial containing 10 µL of RNA (1 µg/µL). Store at -80°C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

SIMPLICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service