Skip to Content
Merck
All Photos(3)

Documents

SCM302

Sigma-Aldrich

Definitive Endoderm Induction Medium

Synonym(s):

Definitive Endoderm Medium, Endoderm Induction Medium, Medium for Endoderm Induction

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.71

form

liquid

packaging

pkg of 50 mL

technique(s)

cell culture | stem cell: suitable

impurities

<2 EU/mL endotoxin

pH

7.0-7.4

application(s)

cell analysis

General description

The definitive endoderm generates the epithelial lining of the respiratory and digestive tracts as well as cell of the thyroid, thymus, lungs, liver and pancreas. The definitive endoderm induction medium (SCM302) is a defined, animal component-free, ready-to-use serum-free medium for efficient differentiation of human pluripotent stem cells (ES/iPSCs) to definitive endoderm (DE) cells.

The 3-day differentiation protocol is simple and has been validated on multiple human iPS cell lines. Differentiated cells express high levels (>85%) of endoderm markers including CXCR4, c-Kit, Sox-17 and FOXA2 and can be further differentiated into specialized endodermal cell lineages. Definitive Endoderm Induction Medium can be used to generate 3D organoids such as hiPS derived colon (Cat. No. SCC300) and lung organoids.

Application

Ready-to-use serum-free media for the efficient differentiation of human pluripotent stem cells (ES/iPSCs) to CXCR4+/c-Kit+/Sox-17+/FOXA2+ definitive endoderm (DE) cells.

Research Category:
Stem Cell Research

Quality

Appearance: Clear liquid/no particulates
Sterility Tested: No growth
Endotoxin: <2 EU/mL
pH: 7.0-7.4
Mycoplasma: Negative
Functional Assay: >85% CXCR4+ c-Kit+ double positive after 3 days of induction.

Storage and Stability

Upon receipt, store at -20°C. When ready to use, thaw overnight at 2-8°C. Once thawed, mix thoroughly and aliquot into smaller volumes (i.e. 10 mL), use immediately and store at 2-8°C for up to 1 week. Do not re-freeze. Unused aliquots may be stored at -20°C until the expiry date.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Protocols

Highly characterized cryopreserved human colonic organoids and a step-by-step organoid culture protocol for epithelial intestinal organoid differentiation from iPS cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service