Skip to Content
Merck
All Photos(1)

Key Documents

SCM081

Sigma-Aldrich

N21 Medium Supplement (50X)

The N21 Medium Supplement is an optimized serum-free supplement developed for isolation & expansion of neurons from mouse & rat hippocampus.

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$657.00

$657.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
10 ML
$657.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.71

$657.00


Estimated to ship on18 May 2025


Request a Bulk Order

Quality Level

form

liquid

manufacturer/tradename

Millipore

technique(s)

cell culture | stem cell: suitable

input

sample type: mouse embryonic stem cell(s)
sample type neural stem cell(s)
sample type induced pluripotent stem cell(s)

shipped in

dry ice

General description

N21 Medium Supplement is an optimized serum-free supplement developed for isolation and expansion of neurons from mouse and rat hippocampus. In addition to its use for long term neuronal survival, it has also been validated for serum-free culture of mouse ES cells and human ES cell derived neural progenitor cells.

Application

The N21 Medium Supplement is an optimized serum-free supplement developed for isolation & expansion of neurons from mouse & rat hippocampus.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service