Skip to Content
Merck
All Photos(6)

Key Documents

SCC494

Sigma-Aldrich

Mouse OP9-DL4 Delta-like 4 Notch Ligand Cell Line

Mouse

Synonym(s):

OP9-DL4;OP9-DL4 cell line;DL4 cell line;Delta-like 4 cell line;Notch ligand cell line

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$1,830.00

$1,830.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 VIAL
$1,830.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.45

$1,830.00


Estimated to ship on29 May 2025


Product Name

Mouse OP9-DL4 Delta-like 4 Notch Ligand Cell Line,

biological source

mouse

Quality Level

packaging

vial of ≥1X10⁶ cells

manufacturer/tradename

Millipore

technique(s)

cell culture | mammalian: suitable
cell differentiation: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

Cell Line Description

OP9-DL4 is a stable bone marrow derived stromal cell line expressing the Notch ligand- Delta like 4 (DL4) ectopically.Notch signaling controls multiple cell-fate decisions. Four Notch receptors (Notch 1- Notch 4) have been identified in mammals. These receptors interact with Jagged or Delta-like (DL) family members of Notch ligands. This is followed by cleavage of the intracellular domain of Notch and its subsequent translocation into the nucleus, where it binds with transcription factors and activates transcription of various downstream target genes. Notch signaling is essential in early T- cell lineage commitments. Bone marrow progenitor cells expressing constitutively-active Notch develop into CD4 and CD8 double positive T cells rather than B cells. They also play an important role in development of CD4+ and CD8+ single positive cells from double positive precursor thymocytes, in addition to development of T-cells with αβ+ TCRs. Of the 4 Notch receptors, Notch1 (N1) receptor signaling has been shown to be sufficient for T cell development. The delta like ligands are physiologically relevant N1 ligands. DL1 interacts with both N1 and Notch 2 (N2) to induce T- cell lineage commitment. However, DL4 interacts specifically only with N1 and supports T cell commitment and maturation both in vitro and in vivo. Moreover, results from previous binding studies show that binding between DL1 and N1 is weak as opposed to the stronger binding between DL4 and N1. Of the DL1 and DL4 delta like ligands, DL4 exhibits greater capacity to activate the Notch pathway in hematopoietic progenitor cells. SourceOP9-DL4 cells were genetically modified from OP9 stromal cells derived from mouse bone marrow.References1.Immunity 2002, 17(6): 749-756.2.J Orthop Sci. 2005, 10(6): 589-594.3.Curr Opin Immunol. 2007, 19(2): 163-168. 4.Cold Spring Harb Protoc. 2009, 2009(2): pdb.prot5156.

Application

  • Each vial contains > 1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of mouse origin and negative for interspecies contamination from human, rat, Chinese hamster, Golden Syrian hamster, and nonhuman primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination.

Features and Benefits

Mouse OP9-DL4 cells express the Notch ligand Delta-like 4 and have been demonstrated to promote T cell lineage commitment in hematopoietic stem cells. These cells support the differentiation of embryonic or hematopoietic stem cells from fetal liver or bone marrow into T lymphocytes. The OP9-DL4 cell line is an essential tool for elucidating Notch signaling in cellular differentiation and the initiation of inflammatory disease states.

Storage and Stability

OP9-DL4 cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages without significantly affecting cell marker expression and function.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Could you provide information on whether the DL4 expressed by the cells is human or mouse DL4?

    1 answer
    1. The mouse for Product No. SCC494 is the Mouse OP9-DL4 Delta-like 4 Notch Ligand Cell Line.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service