Skip to Content
Merck
All Photos(1)

Key Documents

S7804

Sigma-Aldrich

CpG WIZ E-Cadherin Amplification Kit

The CpG WIZ E-cahedrin Amplification Kit is used for determining the methylation of status of the E-cadherin promoter by methylation-specific PCR (MSP).

Synonym(s):

CD324

Sign Into View Organizational & Contract Pricing

Select a Size

25 ASSAYS
$1,230.00

$1,230.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
25 ASSAYS
$1,230.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.31

$1,230.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

species reactivity

human

manufacturer/tradename

CPGWIZ
Chemicon®

technique(s)

PCR: suitable

NCBI accession no.

UniProt accession no.

application(s)

genomic analysis

shipped in

dry ice

Related Categories

General description

E-cadherin is a cell adhesion protein that suppresses tumor cell invasion and metastasis in tumor models. On mechanism regulating the expression of the E-cadherin gene is methylation of the promoter region. The CpG WIZ E-cahedrin Amplification Kit is used for determining the methylation of status of the E-cadherin promoter by methylation-specific PCR (MSP). The kit contains primers targeted to regions of the promoter where the sequences are most divergent after bisulfite treatment. PCR parameters have been identified so that all primer sets in the kit amplify under the same conditions. Control genomic DNA samples (methylated and unmethylated) for E-cadherin are also included.



Methylation-specific PCR (MSP) is a new technology for sensitive detection of abnormal gene methylation utilizing small amounts of DNA (Herman, 1996 ). This process employs an initial bisulfite reaction to modify the DNA, followed by PCR amplification with specific primers designed to distinguish methylated from unmethylated DNA.

Application

Principles of the Technique

Use of either the CpGenome DNA Modification Kit (Cat. No. S7820) or the CpGENOME Fast DNA Modification Kit (Cat. No. S7824) facilitates the initial bisulfite reactions, while the CpG WIZ E-cadherin Amplification Kit contains the reagents required for the gene-specific PCR amplification reactions.
The CpG WIZ E-cahedrin Amplification Kit is used for determining the methylation of status of the E-cadherin promoter by methylation-specific PCR (MSP).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
CPGWIZ is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service