Skip to Content
Merck
All Photos(1)

Key Documents

S4040

Millipore

Hybrisol® I Hybridization Solution

50% Formamide, 6X SSC

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$827.00

$827.00


Estimated to ship on30 March 2025



Select a Size

Change View
500 ML
$827.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.25

$827.00


Estimated to ship on30 March 2025


Product Name

Hybrisol® I Hybridization Solution, (50% Formamide, 6X SSC),

packaging

pkg of 500 mL

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

Northern blotting: suitable
Southern blotting: suitable

shipped in

ambient

General description

Hybrisol® Hybridization Solutions are pre-made, ready-to-use solutions optimized for sensitive single and multiple gene copy detection using membrane-based hybridization techniques. They are designed to give maximum signal and clean background for hybridizations performed on nylon and nitrocellulose membranes. Hybrisol Solution contains 10% Dextran Sulfate, 1% SDS, 50% Formamide, 6X SSC, Sheared DNA, and Modified Denhardt′s Solution.

Application

All
Before using, preheat at 37°C to 45°C for 10 - 20 minutes to dissolve precipitants and mix thoroughly. Use 1 ml for every 15 cm2 membrane for prehybridization and hybridization. Prehybridize for 60 minutes at 42°C to 45°C. Hybridize overnight at 45°C to 52°C. Prehybridization and hybridization temperatures may have to be determined empirically. Refer to the Nomograph located under the "Protocols" link at S4040 product specific page at www.chemicon.com for hybridization temperatures with various stringencies guidelines.

Physical form

Liquid Hybrisol I Solution contains 10% Dextran Sulfate, 1% SDS, 50% Formamide, 6X SSC, Sheared DNA, and Modified

Denhardt′s Solution.

Storage and Stability

Store at room temperature tightly sealed when not in use. Solution can be stored refrigerated however it should be warmed to at least room temperature prior to use dilution for best results. Precipitates may form when the solution is cooled.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
HYBRISOL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A general method for the isolation of RNA complementary to DNA.
BOLTON, E T and McCARTHY, B J
Proceedings of the National Academy of Sciences of the USA, 48, 1390-1397 (1962)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service