Skip to Content
Merck
All Photos(1)

Key Documents

QIA88

Millipore

ProteoExtract® Cytosol/Mitochondria Fractionation Kit

Synonym(s):

Cellular Fractionation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500

usage

sufficient for 100 extractions

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

DNA extraction: suitable
RNA extraction: suitable
nuclear extraction: suitable
protein extraction: suitable

input

sample type cells

shipped in

dry ice

storage temp.

−20°C

General description

Note: 1 Test = reagents sufficient for processing 5 x 107 cells.
This kit provides a unique combination of reagents useful for the isolation of a highly enriched mitochondrial fraction from the cytosol. The enriched fractions can be used to study factors of interest using Western blotting, ELISA, or other assays.

Components

Mitochondria Extraction Buffer, Cytosol Extraction Buffer, Protease Inhibitor Cocktail, Reducing Reagent, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Principle

The ProteoExtract Cytosol/Mitochondria Fractionation Kit is designed for extraction of mammalian cells to yield enriched mitochondrial and cytosolic fractions for use in downstream analyses, such as Western blotting.

Preparation Note

• Protease Inhibitor Cocktail: Reconstitute the lyophilized Protease Inhibitor Cocktail by adding 250 µl DMSO; mix well before use. Dispense into aliquots and store at -20°C.• 1X Cytosol Extraction Buffer Mix: Prepare 1X Cytosol Extraction Buffer by diluting the Cytosol Extraction Buffer 1:4 with ddH2O. Prepare only the amount needed for the assay (5 x 107 cells require 1 ml 1X Cytosol Extraction Buffer Mix). Just prior to use add 2 µl Protease Inhibitor Cocktail and 1 µl DTT to 1 ml 1X Cytosol Extraction Buffer.• Mitochondria Extraction Buffer Mix: Prepare only the amount needed for the assay (5 x 107 pelleted cells require 100 µl Mitochondria Extraction Buffer Mix). Just prior to use, add 2 µl Protease Inhibitor Cocktail and 1 µl DTT to 1 ml of Mitochondria Extraction Buffer.

Storage and Stability

Upon arrival store the entire contents of the kit at -20°C. Following initial use, store the buffers at 4°C and the DTT and Protease Inhibitor Cocktail at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEOEXTRACT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiuyun Luo et al.
Oncology research, 28(4), 331-344 (2020-02-26)
Despite therapeutic advances, the effective treatment for relapsed or refractory diffuse large B-cell lymphoma (DLBCL) remains a major clinical challenge. Evasion of apoptosis through upregulating antiapoptotic B-cell lymphoma-2 (BCL-2) family members and p53 inactivation, and abnormal activation of B-cell receptor
Ga Bin Park et al.
Journal of immunology (Baltimore, Md. : 1950), 185(12), 7274-7284 (2010-11-17)
CD70 is expressed in normal activated immune cells as well as in several types of tumors. It has been established that anti-CD70 mAb induces complement-dependent death of CD70(+) tumor cells, but how anti-CD70 mAb affects the intrinsic signaling is poorly
Dominique J Favreau et al.
Journal of virology, 86(1), 81-93 (2011-10-21)
Human coronaviruses (HCoV) are recognized respiratory pathogens. Some HCoV strains, including HCoV-OC43, can invade the central nervous system, where they infect neurons, with unclear consequences. We have previously reported that HCoV-OC43 infection of human neurons activates the unfolded-protein response and
Hao Lu et al.
BMC veterinary research, 9, 217-217 (2013-10-24)
Locoweeds cause significant livestock poisoning and economic loss all over the world. Animals can develop locoism, a chronic neurological disease, after grazing on locoweeds. Oxytropis kansuensis is a variety of locoweed that contains swainsonine as its main toxic ingredient. The
Adaikkappan Periyakaruppan et al.
Archives of toxicology, 83(6), 595-600 (2008-12-20)
Uranium is a naturally occurring radioactive material present everywhere in the environment. It is toxic because of its chemical or radioactive properties. Uranium enters environment mainly from mines and industry and cause threat to human health by accumulating in lungs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service