Skip to Content
Merck
All Photos(1)

Key Documents

PX2014

Supelco

Pyridine

OmniSolv®

Synonym(s):

Pyridine, Azabenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-809-9
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
115 °C/1013 hPa
vapor pressure:
26.7 hPa ( 25 °C)
Pricing and availability is not currently available.

vapor pressure

26.7 hPa ( 25 °C)

Quality Level

product line

OmniSolv®

Assay

≥99% (GC)

form

liquid

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.05% Water (H2O)

evapn. residue

≤4 ppm

color

APHA: ≤10, at time of manufacture

pH

8.81 (20 °C in H2O)

bp

115 °C/1013 hPa

mp

-42 °C

transition temp

flash point 17 °C

density

0.982 g/cm3 at 20 °C

UV cutoff

≤330 nm

UV absorption

λ: 330 nm Amax: ≤1.00
λ: 340 nm Amax: ≤0.30
λ: 350 nm Amax: ≤0.10
λ: 400 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

SMILES string

n1ccccc1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Application

OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.0% minColor (APHA) at time of manufacture10 maxEvaporation residue4ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 330 nm1.00AU maxUV Abs. at 340 nm0.30AU maxUV Abs. at 350 nm0.10AU maxUV Abs. at 400 nm0.01AU maxUV Cut-off330nm maxWater (H2O)0.05% max
Evaporation Number: 12.7

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service