Skip to Content
Merck
All Photos(3)

Key Documents

PD2004705

Millipore

Petri dish

sterile, Single use, diam. 47 mm, pkg of 600 units ((4x150)), suitable for microbiology

Sign Into View Organizational & Contract Pricing

Select a Size

600 UNITS
$473.00

$473.00


Check Cart for Availability

Request more information

Select a Size

Change View
600 UNITS
$473.00

About This Item

UNSPSC Code:
41122101
eCl@ss:
32030209
NACRES:
NB.14

$473.00


Check Cart for Availability

Request more information

material

cellulose pad(s)
clear device
polystyrene

Quality Level

sterility

sterile

packaging

pkg of 600 units ((4x150))

diam.

47 mm

color

clear

input

sample type dairy
sample type soft drink
sample type beverage(s)
sample type bottled water
sample type food(s)
sample type beer
sample type wine
sample type cider

application(s)

bioburden testing
(Beverages, Water)

microbiology

compatibility

(Membrane filtration)

shipped in

ambient

General description

Petri dishes (47 mm) are single-use, disposable plastic units for culturing micro-organisms captured through membrane filtration. The dishes have squared-off edges for one-handed opening, are easily stackable, and have snug-fitting lids to prevent dehydration of media.

Application

Petri dish is used to culture micro-organisms captured on a 47 mm filter through membrane filtration. It is used for testing in the Dairy, Beer, Cider, Soft Drink, Wine, and Bottled Water industries.

related product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service