Skip to Content
Merck
All Photos(1)

Key Documents

PC709

Sigma-Aldrich

Anti-MYH (33-51) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-MutY Homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$772.00

$772.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$772.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$772.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUTYH(4595)

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~52-57 kDa isoforms of MYH.
Recognizes the MYH protein and several of its approximately 10 isoforms in PC12, Jurkat, and HeLa cells.
This Anti-MYH (33-51) Rabbit pAb is validated for use in Immunoblotting, Immunofluorescence, Paraffin Sections for the detection of MYH (33-51).

Immunogen

Human
a synthetic peptide (QEGRQKHAKNNSQAKPSAC) corresponding to amino acids 33-51 of human MYH

Application

Immunoblotting (1:1500)

Immunofluorescence (1:500)

Paraffin Sections (see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
PC12, Jurkat, or HeLa cells

Other Notes

Detects MYH and several of its approximately ten isoforms. In Jurkat cells the ~52 kDa and ~53 kDa variants were found in the nuclear fraction while the ~57 kDa protein is found in the mitochondria. Antibody should be titrated for optimal results in individual systems.
Wang, G., et al. 2000. Nucleic Acids Res.28, 2135.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service