Key Documents
PA25/111
Millipore
Pneumatic Plastic Actuator for NOVASEPTIC® Valve, Normally Closed (NC)
About This Item
UNSPSC Code:
31251504
Pricing and availability is not currently available. Contact Technical Service
material
plastic cover (POM C)
plastic indicator (POM C)
stainless steel cover
Quality Level
Agency
meets EN ASTM
meets EN Stainless Steel EN 1.4305
parameter
0-70 °C temp. range
1.5 kg pressure range (3.31 lb)
roughness average (RA)
1.6 μm ((63 μin) Electropolished)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Actuator is packaged in a closed box.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany