Skip to Content
Merck
All Photos(1)

Documents

OX0270

Supelco

Oxalic Acid, Dihydrate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Oxalic Acid, Dihydrate, Ethanedioic Acid, Dihydrate, Ethanedioic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H2O4 · 2H2O
CAS Number:
Molecular Weight:
126.07
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-634-3

Agency

meets USP/NF

Quality Level

vapor pressure

0.000312 hPa ( 25 °C)

Assay

99.5-102.5%

form

solid

impurities

≤0.005% Insoluble matter

ign. residue

≤0.01%

pH

1.5 (10 g/L in H2O)

bp

149-160 °C/1013 hPa (decomposition)

mp

98-100 °C (Elimination of water of crystallisation)

transition temp

flash point 157 °C (decomposition)

solubility

>100 g/L

density

1.65 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.001%
Fe: ≤2 ppm
N: ≤0.001%
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

InChI

1S/C2H2O4.2H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H2

InChI key

GEVPUGOOGXGPIO-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay: 99.5-102.5%
Calcium (Ca): 0.001% max
Chloride (Cl): 0.002% max
Heavy Metals (by ICP-OES): 5ppm max
Insoluble matter: 0.005% max
Iron (Fe): 2ppm max
Nitrogen compounds (as N): 0.001% max
Residue after ignition: 0.01% max
Substances darkened by hot sulfuric acid: To pass test
Sulfate (SO4): 0.005% max

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service