Skip to Content
Merck
All Photos(1)

Key Documents

OP97

Sigma-Aldrich

Anti-mTOR/FRAP (Ab-2) Mouse mAb (22C2)

liquid, clone 22C2, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

22C2, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MTOR(2475)

General description

Purified mouse monoclonal antibody generated by immunizing mice with the specified immunogen and fusing splenocytes with SpAG8 mouse myeloma cells. Recognizes the ~290 kDa mTOR/FRAP protein.
Recognizes the ~290 kDa mTOR protein in HEK293 and Jurkat cells.
This Anti-mTOR/FRAP (Ab-2) Mouse mAb (22C2) is validated for use in Immunoblotting, Immunoprecipitation for the detection of mTOR/FRAP (Ab-2).

Immunogen

Human
a synthetic peptide corresponding to amino acids 230-240 of human TOR

Application


Immunoblotting (1.7 g/ml, see application references)
Immunoprecipitation (1-10 g/ml, see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Positive Control
Jurkat or HeLa cells

Other Notes

Kimura N et al. 2003. Genes Cells8, 65.
Sekulic A et al. 2000. Cancer Res.60, 3504.
Hosoi, H. 1999. Cancer Res.59, 886.
Alarcon, C.M., et al. 1996. Genes Dev.10, 279.
Freeman, K., and Livi, G.P. 1996. Gene172, 143.
Hosoi, H., et al. 1996. AACR 87th Ann. Mtg. Abstract 3445.
Cardenas, M.E., and Heitman, J. 1995. EMBO J.14, 5892.
Lorenz, M.C., and Heitman, J. 1995. J. Biol. Chem.270, 27531.
Sabatini, D.M., et al. 1995. J. Biol. Chem.270, 20875.
Zheng, X.F., et al. 1995. Cell82, 121.
Brown, E.J., et al. 1994. Nature369, 756.
Sabatini, D.M., et al. 1994. Cell78, 35.
Stan, R., et al. 1994. J. Biol. Chem.269, 32027.
Kunz, J., et al. 1993. Cell73, 585.
Heitman, J., et al. 1991. Science253, 905.
The immunogen sequence is 100% conserved in mouse and rat, but cross-reactivity has not been tested. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuliana Gatti et al.
Biomedicines, 9(10) (2021-10-24)
Nuclear lamina components have long been regarded as scaffolding proteins, forming a dense fibrillar structure necessary for the maintenance of the nucleus shape in all the animal kingdom. More recently, mutations, aberrant localisation and deregulation of these proteins have been
Kuppusamy Balamurugan et al.
Nature communications, 4, 1662-1662 (2013-04-12)
Toll-like receptor 4 (Tlr4) has a pivotal role in innate immune responses, and the transcription factor CCAAT/enhancer binding protein delta (C/EBPδ, Cebpd) is a Tlr4-induced gene. Here we identify a positive feedback loop in which C/EBPδ activates Tlr4 gene expression
V Felzen et al.
Cell death & disease, 6, e1812-e1812 (2015-07-15)
Breast cancer is a heterogeneous disease and approximately 70% of newly diagnosed breast cancers are estrogen receptor (ER) positive. Out of the two ER types, α and β, ERα is the only ER that is detectable by immunohistochemistry in breast
Debapriya Chakraborty et al.
Redox biology, 24, 101181-101181 (2019-04-09)
Oxidative stress and a disturbed cellular protein homeostasis (proteostasis) belong to the most important hallmarks of aging and of neurodegenerative disorders. The proteasomal and autophagic-lysosomal degradation pathways are key measures to maintain proteostasis. Here, we report that hippocampal cells selected

Articles

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service