Skip to Content
Merck
All Photos(3)

Key Documents

NATQ04725

Millipore

Polyacrylamide Membrane Filter, 0.4 μm Pore Size

Natrix® Q, filter diam. 47 mm, hydrophilic

Synonym(s):

Hydrogel, Natrix, Natrix® Q Membrane Filter, Quaternary Amine Membrane

Sign Into View Organizational & Contract Pricing

Select a Size

25 EACH
$796.00

$796.00


Check Cart for Availability


Select a Size

Change View
25 EACH
$796.00

About This Item

UNSPSC Code:
40161507
NACRES:
NB.21

$796.00


Check Cart for Availability

material

plain filter
polyacrylamide (Crosslinked Polyacrylamide with Quaternary Amine functionality)
white filter

Quality Level

feature

hydrophilic

manufacturer/tradename

Millipore
Natrix® Q

parameter

6-10 mL/min water flow rate (at 15 psi)

diam.

47 mm

filter diam.

47 mm

thickness

200-300 μm , dry thickness

pore size

0.4 μm pore size (nominal)

Looking for similar products? Visit Product Comparison Guide

General description

The unique Natrix® Q chromatography membrane is a porous polyacrylamide hydrogel containing a high density of pendant quaternary ammonium (Q) binding groups that are directly polymerized within an inert, macroporous membrane scaffold.
Filter Code: NATQ

Application

  • Biomolecule purification
  • Impurity or host cell protein removal
  • Bacteriophage purification
  • Viral vector isolation/purification
  • Nucleic acid binding, including eDNA
  • Isolation of virus-like particles in wastewater

Other Notes

Directions for Use:

  • Organism Retention: Microorganism
  • Mode of Action: Charge Exclusion/ Charge Affinity
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Filtration of a liquid through a membrane with a positively surface charge creating an affinity for negatively charged biological contaminants, including bacteria, viruses, cell particulate and antibodies.
  • Storage Statement: Store in a dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

NATRIX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

See case study examples of how to optimize chromatographic purification of plasmid DNA for Biopharmaceutical Applications.

Free eBook outlines plasmid DNA manufacturing process and downstream purification challenges for various applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service