Skip to Content
Merck
All Photos(4)

Key Documents

MZHAWG101

Millipore

Microfil® with EZ-Pak®

Microfil® with EZ-Pak®

sample volume 100 mL, ready-to-use, mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, white filter, gridded filter

Synonym(s):

Membrane filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24
Pricing and availability is not currently available. Contact Technical Service

material

gridded filter
mixed cellulose esters (MCE)
polypropylene
white filter

Agency

ISO 7704

description

General microbiological analysis

sterility

sterile; ethylene oxide treated

feature

easily wetted with water
hydrophilic
ready-to-use

manufacturer/tradename

Microfil® with EZ-Pak®

parameter

100 mL sample volume

filter diam.

47 mm

funnel capacity

100 mL

matrix

MF-Millipore

pore size

0.45 μm pore size

input

sample type wine
sample type beverage(s)
sample type cider
sample type soft drink
sample type cosmetics
sample type beer
sample type food(s)
sample type bottled water
sample(s) (environmental)
sample type sport drink

suitability

suitable for microbiology

application(s)

bioburden testing
clinical
cosmetics
environmental
environmental
food and beverages
industrial qc

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Microfil® with EZ-Pak® is an easy to use, fast, and accurate system for the routine microbiological analysis of beverages and drinking water. For the ultimate convenience and efficiency, combine the EZ-Pak® system with the Microfil® filtration unit. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. These ready-to-use 100 mL Microfil® funnels that come with the 0.45 μm pore size, white gridded EZ-Pak® filter system eliminate the need for washing and sterilizing after each test thus making the equipment handling easier and efficient to trap bacteria. The push-fit design of the Microfil® funnels seals tightly to manifold supports without the need for clamps or O-rings. Unlike with conventional filter holders, incomplete sealing at the membrane/funnel interface is eliminated resulting in consistent recovery of organisms.

Application

Microfil® with EZ-Pak® filters find application in Clinical, Cosmetics, Environmental, Industrial QC, Pharmaceutical, Food and Beverage industries. Microbial analysis of beer, bottled water, cider, sports drink, and wine by membrane filtration method

Features and Benefits

  • Convenient and easy to use
  • Sterilized and ready-to-use device- saves time and expense
  • Reduce laboratory workload
  • Faster filtration rate due to the larger surface area

Principle

Vacuum

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service