MX0075
Magnesium sulfate
anhydrous powder Meets Reagent Specifications for testing USP/NF monographs GR
Synonym(s):
Magnesium Sulfate, Anhydrous, Sulfuric Acid Magnesium Salt, Epsom salts
About This Item
Recommended Products
grade
reagent
Quality Level
Assay
≥98.0% (complexometric)
form
powder
loss
≤2% loss on ignition
color
white
pH
7.9 (25 °C, 50 g/L in H2O)
mp
1124 °C (Lit.)
solubility
300 g/L
anion traces
chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.005%
cation traces
As: ≤0.001%
Ca: ≤0.04%
Fe: ≤0.001%
Mn: ≤0.001%
NH4+: ≤0.005%
heavy metals (as Pb): ≤0.001%
shipped in
ambient
storage temp.
room temp
SMILES string
[Mg+2].[S](=O)(=O)([O-])[O-]
InChI
1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
InChI key
CSNNHWWHGAXBCP-UHFFFAOYSA-L
Related Categories
Application
- Determination of pesticide residues in soybeans using QuEChERS followed by deep eutectic solvent-based DLLME preconcentration prior to gas chromatography-mass spectrometry analysis.: This study highlights a novel application of magnesium sulfate in pesticide residue analysis, employing it within the QuEChERS method to enhance preconcentration and detection capabilities in agricultural chemistry (Petrarca et al., 2024).
Analysis Note
Arsenic (As): 0.001% max
Assay (complexometric): 98.0% min
Calcium (Ca): 0.04% max
Chloride (Cl): 0.001% max
Color: White
Form: Powder
Heavy metals (as Pb): 0.001% max
Iron (Fe): 0.001% max
Loss on ignition: 2.0% max
Manganese (Mn): 0.001% max
Nitrate (NO3): 0.005% max
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service