Skip to Content
Merck
All Photos(3)

Key Documents

MTRACEMM1

Millipore

M-TRACE® Mobile Computer Mount

Synonym(s):

Mobile computer mount

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
43210000
NACRES:
JA.12
Pricing and availability is not currently available. Contact Technical Service

description

For use with M-TRACE™ Touch MobileComputer

packaging

pack of -1 unit

application(s)

pharmaceutical
sterility testing

General description

The M-Trace® Software & Mobile App is your 21 CFR Part 11 compliant Digital QC companion for sterility testing and other regulated processes, ensuring trusted and efficient data recording. Click here to get more details on the overall M-Trace® solution.

M-TRACE® Mobile Computer Mount is an accessory to conveniently mount the M-TRACE® Touch Mobile Computer (MTRACEMC1 ) outside the isolator/LFH pane. The mobile computer mount facilitates hands-free scanning and registration of the information on consumables while the operator′s hands are busy and occupied in the isolator. The M-TRACE® Mobile Computer Mount is clean room friendly and is compliant with the QC sterility testing environment requirements.

Application

M-TRACE® Mobile Computer Mount is used to mount the M-TRACE® Touch Mobile Computer outside the isolator/LFH pane during sterility testing and other quality control workflows.

Legal Information

M-TRACE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

M-Trace® electronic test record software ensures full traceability for your sterility testing data and elevates your Steritest® Symbio pump to a whole new level. M-Trace® software reduces handling errors, ensures compliance, and boosts productivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service