Skip to Content
Merck
All Photos(1)

Key Documents

MSLM10001

Millipore

MilliSnap system large white adapter for incubation on liquid media cassette

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$104.00

$104.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EA
$104.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$104.00


Check Cart for Availability

Request more information

manufacturer/tradename

MilliSnap®

Quality Level

application(s)

bioburden testing
pharmaceutical
water monitoring

shipped in

ambient

General description

Designed to simplify your workflow, the MilliSnap plug and play system is a hands-free method for separating your Milliflex funnel and attaching the membrane to the Millipore media cassettes. The unique convex shape of the Milliflex membrane combined with the MilliSnap system ensures proper contact between membrane and media.

Legal Information

MILLISNAP is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service