Skip to Content
Merck
All Photos(1)

Key Documents

MSIPN4B10

Millipore

Multiscreen® 96 well Plate, hydrophobic PVDF membrane

pore size 0.45 μm, non-sterile

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$463.00

$463.00


Estimated to ship on16 April 2025



Select a Size

Change View
10 EA
$463.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

$463.00


Estimated to ship on16 April 2025


material

Barex®
PVDF membrane (hydrophobic)
flat bottom wells
titanium dioxide

Quality Level

description

Avidin biotin linkages, DNA-binding proteins, protein binding, PAMPA

sterility

non-sterile

product line

MultiScreen®

feature

hydrophobic
lid

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

technique(s)

parallel artificial membrane permeation assay (PAMPA): suitable

H

14.4 mm

L

128 mm

W

85.5 mm

filtration area

0.28 cm2

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

matrix

Immobilon®-P

pore size

0.45 μm pore size

binding type

high binding surface

shipped in

ambient

Application

DNA-binding Proteins and Protein Binding

Legal Information

Barex is a registered trademark of INEOS Barex AG
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Combining solubility & PAMPA assays streamlines drug permeability testing.

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

Protocols

Guidelines for various bioassays on the MultiScreen system, which makes it possible to carry out an assay from sample incubation to scintillation counting in a 96-well plate.

Related Content

This provides information about the chemical compatibility of MultiScreen Filter Plates with different reagents.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service