Skip to Content
Merck
All Photos(1)

Key Documents

MSDVN6B50

Millipore

Multiscreen® 96 well Plate, hydrophilic PVDF membrane

pore size 0.65 μm, non-sterile

Synonym(s):

96-Well PVDF Plate, Hydrophilic 96-Well Plate, PVDF 96-Well Plate

Sign Into View Organizational & Contract Pricing

Select a Size

50 PKG
$2,100.00

$2,100.00


Check Cart for Availability


Select a Size

Change View
50 PKG
$2,100.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

$2,100.00


Check Cart for Availability

material

Barex®
PVDF membrane (hydrophilic)
flat bottom wells
titanium dioxide

Quality Level

description

Receptor binding assays on whole cells and cell fragments, protein kinase/phosphatase precipitation assays, bead/resin based assays

sterility

non-sterile

product line

MultiScreen®

feature

hydrophilic
lid

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

technique(s)

activity assay: suitable (protein kinase/phosphatase precipitation)
bead-based assay: suitable
ligand binding assay: suitable
receptor binding assay: suitable

H

14.4 mm

L

128 mm

W

85.5 mm

filtration area

0.28 cm2

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

matrix

Durapore®

pore size

0.65 μm pore size

binding type

low binding surface

shipped in

ambient

General description

Filter Code: DV
No matter what application you’re working with, Millipore has a plate developed to meet your specific needs. MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.

HTS Filter Plates (Plate catalogue numbers that start with MS)
- MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics.

- Millipore’s new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability in both background and signal intensities specifically for biochemical screening assays.

Classic Filter Plates (Plate catalogue numbers that start with MA)
- The original multiwell design is valuable for many applications which require immersing the filter in an assay tray. The easily removable underdrain and unique plate design are ideal for assays such as Elispot, PAMPA, cell screening (migration, invasion or chemotaxis) or for topcount detection users.

Accessory – MultiScreenHTS Vacuum Manifold
- The MultiScreenHTS Vacuum manifold was designed specifically to work with MultiScreenHTS filter plates. The combination results in optimal filtration characteristics and seamless automation integration.

Millicell Assay Plates
- In addition to the traditional assay plate format, filter plates are also available with cell culture trays for growing and assaying suspension and adherent cell lines. Cells grown on filter plates differentiate better than those grown on traditional plastic substrates, resulting in more in vivo-like results This filter plate/tray combination can be used for a variety of cellular assays, including, transport, toxicity and screening assays.

Application

Research Category
Protein

Assays

Bead/Resin based assays

General Filtration

Physical form

Color Code: Opaque

Other Notes

Automation Compatible: Yes

Legal Information

Barex is a registered trademark of INEOS Barex AG
Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Lind et al.
Scandinavian journal of immunology, 91(4), e12864-e12864 (2020-02-15)
Narcolepsy type 1 (NT1) is a chronic sleep disorder caused by a specific loss of hypocretin-producing neurons. The incidence of NT1 increased in Sweden, Finland and Norway following Pandemrix®-vaccination, initiated to prevent the 2009 influenza pandemic. The pathogenesis of NT1

Articles

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

Protocols

Guidelines for various bioassays on the MultiScreen system, which makes it possible to carry out an assay from sample incubation to scintillation counting in a 96-well plate.

Related Content

This provides information about the chemical compatibility of MultiScreen Filter Plates with different reagents.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service