Skip to Content
Merck
All Photos(1)

Key Documents

MMGSWG124

Millipore

Milliflex Oasis® Filtration Unit

pore size 0.22 μm, mixed cellulose esters (MCE) membrane, white filter, volume 100 mL

Synonym(s):

Sterile membrane filtration device made of gridded, mixed cellulose esters membrane for bioburden testing

Sign Into View Organizational & Contract Pricing

Select a Size

24 EACH
$249.00

$249.00


Check Cart for Availability

Request more information

Select a Size

Change View
24 EACH
$249.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$249.00


Check Cart for Availability

Request more information

material

gridded filter
mixed cellulose esters (MCE) membrane
styrene-butadiene device
white filter

Quality Level

Agency

EP, USP, JP

sterility

electron beam irradiated

feature

ready-to-use

packaging

pack of 24

manufacturer/tradename

Milliflex Oasis®

capacity

100 mL

filter diam.

49 mm

funnel capacity

100 mL

volume

100 mL

pore size

0.22 μm pore size

suitability

suitable for (suitable for solid cultivation)

application(s)

bioburden testing
cosmetics
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® (System)

Looking for similar products? Visit Product Comparison Guide

General description

MILLIFLEX OASIS® 100 mL funnel is a ready-to-use, e-beam irradiated filtration unit that combines a funnel and a gridded membrane filter in a single device offering a high-end solution for bioburden testing. The filtration unit is made of Styrene butadiene copolymer and the membrane is made of mixed cellulose esters with 0.22 μm pore size, white gridded and 49mm diameter. The MILLIFLEX OASIS® membrane filter possesses almost twice the surface area of standard membranes enabling faster filtration and the funnel can be easily detached from the membrane filter without the use of any tools. The convex shape of the membrane filter after filtration demonstrates its integrity and prevents air bubbles from being trapped when placed on culture media.

The MILLIFLEX® PROTACT feature protects your samples from secondary contamination and makes the membrane-to-agar contact optimal delivering the most reliable results. The patented sealed membrane filter feature prevents the bypass phenomenon often found with other filtration systems ensures robust read-out.

The MILLIFLEX® AZURE feature combines transparent funnel and all-around filling level indicators for an easy filling from any angle. The innovative filtration unit assembly design prevents any leaks or sample by-pass of the membrane to ensures the right sample volume is filtered.

The MILLIFLEX® RACK&STACK feature also includes a locked lid for safe transportation and incubation.

Application

MILLIFLEX OASIS® 100 mL funnel find its application in Bioburden testing, water monitoring and general microbiological analysis of beer, bottled water. They are widely used in pharmaceutical and cosmetic industry.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex Oasis® system monitors bioburden in various samples, increasing lab productivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service