Skip to Content
Merck
All Photos(2)

Key Documents

MGAMMAG-300K

Millipore

MILLIPLEX® Mouse Immunoglobulin Isotyping Magnetic Bead Panel - Isotyping Multiplex Assay

Isotyping Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple immunoglobulins (Ig) in murine serum, plasma and cell culture samples.

Synonym(s):

Luminex® Mouse Isotyping Assay, Millipore Mouse Isotyping Assay, Mouse IgG, IgA, IgM immunoassay, Mouse Isotyoing Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

To order a Milliplex® kit, please search for your analyte of interest.

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

accuracy: 100%
(IgM)

accuracy: 85%
(IgG3)

accuracy: 98%
(IgA)

accuracy: 98%
(IgG2b)

sensitivity: 0.18 ng/mL
(IgG2b)

sensitivity: 0.32 ng/mL
(IgG3)

sensitivity: 0.67 ng/mL
(IgG2a)

sensitivity: 0.7 ng/mL
(IgG1)

sensitivity: 1.1 ng/mL
(IgA)

sensitivity: 1.73 ng/mL
(IgM)

standard curve range: 1-900 ng/mL
(IgG2b)

standard curve range: 2-1,500 ng/mL
(IgG1)

standard curve range: 2-1,500 ng/mL
(IgG3)

standard curve range: 2-1,500 ng/mL
(IgM)

standard curve range: 3-2,000 ng/mL
(IgG2a)

standard curve range: 5-3,000 ng/mL
(IgA)

inter-assay cv: 14-17%
intra-assay cv: 5-10%

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Produced by plasma cells and lymphocytes, immunoglobulins (antibodies) are critically involved in immune response, attaching to antigens, and playing a role in their destruction. Immunoglobulins (Ig) can be classified by isotype, classes that differ in function and antigen response due to structure variability. Six major isotypes have been identified in placental mammals: IgM, IgG1, IgG2a, IgG2b, IgG3 and IgA – all found in normal individuals. Immunoglobulin-deficiency disorders, such as autoimmune disease, some GI conditions, and malignancies, are characterized by specific isotype deficiencies or varying concentrations of one or more isotypes. Disease states can range from the absence of one isotype class or subclass to a total deficiency of immunoglobulin classes.

MILLIPLEX® Mouse Isotyping Bead Panel, measuring mouse IgG subclasses (1, 2a, 2b, 3), IgM, and IgA, has been designed to enable you to measure both classes and subclasses accurately – all in one well. In addition, this panel allows you to choose only those immunoglobulins you need for your experiments.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Immune Response

Application

  • Analytes: IgG1, IgG2a, IgG2b, IgG3, IgA, IgM
  • Recommended Sample type: Serum, plasma, or tissue/cell lysate and culture supernatant
  • Recommended Sample dilution: 1:50,000 diluted plasma or serum, or cell culture supernatant diluted to approximately 1μg/mL Ig well.
  • Assay Run Time: One day
  • Research Category: Immunology and Inflammation, Isotyping

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Linkage

Replaces: MGAM-300

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Common tolerance mechanisms, but distinct cross-reactivities associated with gp41 and lipids, limit production of HIV-1 broad neutralizing antibodies 2F5 and 4E10.
Chen, Y; Zhang, J; Hwang, KK; Bouton-Verville, H; Xia, SM; Newman, A; Ouyang, YB; Haynes et al.
Journal of immunology (Baltimore, Md. : 1950) (1950)
Tao Huang et al.
JCI insight, 1(7), e86689-e86689 (2016-10-05)
Eosinophilic inflammation and Th2 cytokine production are central to the pathogenesis of asthma. Agents that target either eosinophils or single Th2 cytokines have shown benefits in subsets of biomarker-positive patients. More broadly effective treatment or disease-modifying effects may be achieved
Mara A Correa et al.
BioMed research international, 2018, 1267038-1267038 (2018-11-08)
Mouse lines selected for maximal (AIRmax) or minimal acute inflammatory reaction (AIRmin) were used to characterize the immune response and the influence of genetic background during pristane-induced arthritis (PIA). Susceptible AIRmax mice demonstrated exacerbated cellular profiles during PIA, with intense
Sungwoo Chei et al.
Frontiers in nutrition, 7, 583186-583186 (2020-12-18)
Anti-aging research suggests that immunosenescent cells can play deleterious roles in the immune system. Here, young (2 months old) and old (14 months old) C57BL/6 mice received a daily oral dose (100 or 750 mg/kg/day) of acid-hydrolyzed silk peptide (SP)
Silke E Lindner et al.
ImmunoHorizons, 4(5), 274-281 (2020-05-22)
Rho family GTPases are critical for normal B cell development and function, and their activity is regulated by a large and complex network of guanine nucleotide exchange factors (GEFs) and GTPase-activating proteins (GAPs). However, the role of GAPs in B

Related Content

Get even more out of your samples with this Luminex® technology-based mouse cytokine assay – the MILLIPLEX® Mouse Cytokine Expanded Panel 1.

Immunology multiplex assays, like MILLIPLEX® assays, allow researchers to investigate more immune biomarkers simultaneously, saving time and resources. This is especially critical to immunology research, where numerous analytes play a role in different pathways.

Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service