Skip to Content
Merck
All Photos(5)

Key Documents

MATPAM3CSK4

Millipore

PAM3CSK4 Non-Endotoxin Pyrogen (NEP) Control

for use with PyroMAT®

Synonym(s):

Lipopeptide, Triacylated Lipopeptide

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,370.00

$1,370.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EACH
$1,370.00

About This Item

UNSPSC Code:
41116129
NACRES:
NA.74

$1,370.00


Check Cart for Availability

Request more information

Agency

EP
JP 4.06
USP

Quality Level

form

lyophilized

manufacturer/tradename

PyroMAT®

application(s)

pharmaceutical
pyrogen testing

compatibility

for use with PyroMAT®

shipped in

ambient

storage temp.

2-8°C

suitability

pyrogenic bacteria

General description

PAM3CSK4 is a non-endotoxin pyrogen which will serve as a positive control during pyrogen testing with PyroMAT® system. Non- Endotoxin Pyrogen (NEP) Control PAM3CSK4 is a synthetic synthetic triacylated lipopeptide, supplied in a lyophilized form. PAM3CSK4 Non- Endotoxin Pyrogen (NEP) Control activates TLR1/TLR2 receptor of monocytes and is used as a positive control for PyroMAT® system. The response to pyrogenic substances is determined by measuring the Interleukin-6 (IL-6) produced by the monocytes using ELISA microplate.

Application

PAM3CSK4 control can be used as a positive control with the PyroMAT® System. It is to be used for Product Specific Validation (PSV) and routine testing to show that the system can detect non-endotoxin pyrogen contaminants in pharmaceutical products.

Legal Information

PYROMAT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service