Skip to Content
Merck
All Photos(1)

Key Documents

MANMN6010

Millipore

Multiscreen® 96 well Plate, mesh nylon membrane

pore size 60.0 μm, non-sterile

Synonym(s):

Mesh Nylon 96-Well Plate, Nylon Membrane Plate

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$1,200.00

$1,200.00


Estimated to ship on08 July 2025FromTruganina



Select a Size

Change View
10 EA
$1,200.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

$1,200.00


Estimated to ship on08 July 2025FromTruganina


material

flat bottom wells
mesh Nylon membrane
polystyrene

Quality Level

description

Whole Organism Screening

sterility

non-sterile

product line

MultiScreen®

feature

lid

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

filtration area

0.28 cm2

plate size

96 wells

well maximum volume

300 μL

working volume

50-250 μL

pore size

60.0 μm pore size

shipped in

ambient

General description

MultiScreen-Mesh plate with receiver plate provides a complete system for culturing, testing and reading assay results in a one-step protocol. This system is ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model.



MultiScreen-Mesh is optimized for compound and target identification in a 96-well format. The 96-well nylon mesh plate and 96-well tray fit together to provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms. Select from a range of mesh pore sizes to appropriate for your application. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms. The plates are optically clear for use with an imaging microscope or other microscopic analysis. Plates can be coated with a thin layer of agar if necessary.



MultiScreen-Mesh plates have also been shown to be useful as high throughput cell strainers prior to cellular analysis in a FACS analyzer. To dissociate clumps of cells prior to loading plate into FACS instrument, run cell suspensions through the mesh plates either by gravity flow or via gentle centrifugation.



This filter plate provides a complete system for culturing, testing and reading assay results in a one-step protocol. The system is ideal for target screening and other applications that evaluate the effect of new compounds on whole, multicellular organisms. The MultiScreen®-Mesh filter plate is optimized for compound and target identification in a 96-well format. The filter plate and 96-well tray fit together to provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.



Features & Benefits:

- Multiple configurations and applications

- Chemical compatibility

- Automation-compatible

- Superior vacuum filtration and filtrate collection

- Plates are optically clear for use with an imaging microscope or other microscopic analysis



Applications

- Whole Organism Screening

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service