Skip to Content
Merck
All Photos(1)

Key Documents

MAGNA0024

Sigma-Aldrich

MAGNA CHIP® Controls, Proteinase K and Protease Inhibitor Cocktails

Sign Into View Organizational & Contract Pricing

Select a Size

MAGNA0024
$472.00

$472.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
MAGNA0024
$472.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.84

$472.00


Estimated to ship on19 May 2025


Request a Bulk Order

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service