Skip to Content
Merck
All Photos(1)

Key Documents

MABX5523

Sigma-Aldrich

Anti-PSA (10mg) KC

from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$276.00
10 MG
$2,760.00

$276.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$276.00
10 MG
$2,760.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$276.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

mouse

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

Gene Information

human ... KLK3(354)

General description

Prostate Specific Antigen (PSA) is a tumor associated antigen with a molecular weight of 34 kD. PSA is found in normal prostatic epithelial lining cells of the ducts and acini, as well as in primary and metastatic prostate carcinoma cells. Measurement of the level of PSA in male blood can help to support the diagnosis of prostate cancer. In addition to prostate cancer, a number of benign (not cancerous) conditions can cause male PSA levels to rise.

Specificity

Specific for free PSA and PSA-ACT (PSA-alpha 1-antichymotrypsin) complex. No cross reactivity with human kallikrein and ACT

Immunogen

corresponding to Human Prostate specific antigen.

Application

Detect Prostate specific antigen using this mouse monoclonal Anti-PSA (10mg) KC, Cat. No. MABX5523-10KC, validated for use in Lateral Flow Assay and ELISA.
Research Category
Apoptosis & Cancer
Suggested antibody pairs MABX5523-1KC and MABX5532-1KC

Quality

Tested

Physical form

Format: Purified
Ion-exchange chromatography
Purified mouse monoclonal in buffer containing 50 mM NaCl, 10 mM PBS, pH 8.0, 0.1 % sodium azide (NaN₃).

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service