Skip to Content
Merck
All Photos(1)

Key Documents

MABT75

Sigma-Aldrich

Anti-Titin (NT/Z-domain) Antibody, clone 17D11.1

clone 17D11.1, from mouse

Synonym(s):

Titin, Connectin, Rhabdomyosarcoma antigen MU-RMS-40.14

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$638.00

$638.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$638.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$638.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

17D11.1, monoclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TTN(7273)

General description

Titin is a large complex protein containing over one hundred IgG-like and fibronectin type-III domains, in addition to multiple Kelch, RCC1, TPR, and WD repeats. It is a member of the CAMK Ser/Thr protein kinase family and also possesses a protein kinase domain, which is involved in controlling gene expression and protein turnover in muscle tissues. Titin also functions as a sensor to mediate appropriate cellular responses to mechanical stress. In striated muscle tissue, titin regulates the length of the sarcomere by integrating myosin and actin filaments at the Z-disk and M-band, and facilitates the elasticity of the sarcomere. Titin also regulates sarcomere assembly and functions as central hub in signal transduction pathways. Defects in Titin may contribute to various forms of cardiomyopathy.

Immunogen

GST-tagged recombinant protein corresponding to human Titin.

Application

Anti-Titin (N-terminal/Z-domain) Antibody, clone 17D11.1 is an antibody against Titin (N-terminal/Z-domain) for use in Western Blotting, IHC(P).
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Immunohistochemistry in human and mouse cardiac muscle tissues.

Immunohistochemistry Analysis: 1:1,000 dilution of this antibody detected Titin in human and mouse cardiac muscle tissues.

Target description

38 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human and mouse cardiac muscle tissues

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Grazia Volpe et al.
Foods (Basel, Switzerland), 8(4) (2019-04-04)
In this study different methods were used to evaluate the effectiveness of a carrageenan coating and carrageenan coating incorporating lemon essential oil (ELO) in preserving the physicochemical and olfactory characteristics of trout fillets stored at 4 °C up to 12

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service