Skip to Content
Merck
All Photos(1)

Key Documents

MABT546

Sigma-Aldrich

Anti-Troponin I Antibody, clone 5C11.1

clone 5C11.1, from mouse

Synonym(s):

Cardiac Troponin I

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$684.00

$684.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$684.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$684.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5C11.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

chicken (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TNNI3(7137)

General description

Troponin I, cardiac muscle (UniProt: P19429; also known as Cardiac troponin I) is encoded by the TNNI3 (also known as TNNC1) gene (Gene ID: 7137) in human. Troponin is a complex that regulates the contraction of skeletal muscle, It consists of 3 subunits known as C, T, and I, which are located periodically along the thin filament of the myofibrils. Troponin C (TnC) binds calcium ions, Troponin I is the inhibitory subunit of troponin that inhibits actomyosin ATPase and Troponin T binds to tropomyosin. It also confers calcium-sensitivity to actomyosin ATPase. Amino acids region 32-79 of Troponin I is involved in binding to TnC and region 129-149 is involved in binding to both TnC and actin. It is thought that troponin I acts as a switch, by moving away from actin toward troponin C, following Ca2+ binding to TnC. Cardiac muscle Troponin I has 31 amino acid residues on its N-terminal that are not present in the skeletal muscle form. Hence, cardiac specificity of this isoform is shown to offer accuracy in diagnosis in patients with acute or chronic skeletal muscle injury. Mutation in TNNI3 gene are reported to result in multiple types of cardiomyopathies that can result in premature death. (Ref.: Li, H-C., and Fajer, PG (1998). Biochemistry 37(19); 6628-6635).

Specificity

Clone 5C11.1 specifically detects Tropinin I in multiple species. It targets an epitope with in 14 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human Troponin I.

Application

Anti-Troponin I, clone 5C11.1, Cat. No. MABT546, is a mouse monoclonal antibody that detects Troponin I and has been tested for use in Western Blotting.

Quality

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Troponin I in human skeletal muscle tissue lysate.

Target description

~22 kDa observed; 24.00 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service