MABT535
Anti-Desmin Antibody, clone 4C12.1
clone 4C12.1, from mouse
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
4C12.1, monoclonal
species reactivity
mouse, rat, human
packaging
antibody small pack of 25 μg
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG2bκ
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... DES(1674)
General description
In adult striated muscle desmin forms a fibrous network connecting myofibrils to each other and to the plasma membrane from the periphery of the Z-line structures. It may also act as a sarcomeric microtubule-anchoring protein and specifically associate with detyrosinated tubulin-alpha chains, leading to buckled microtubules and mechanical resistance to contraction. Desmin is particularly abundant at the myotendinous junctions and at the neuromuscular junctions of skeletal muscle. Mammalian cardiac muscle cells contain higher amounts of desmin (up to 2% of total protein) compared to skeletal muscle cells (0.35% of total protein). Mutations in DES gene have been linked to familial cardiac and skeletal myopathy characterized by skeletal muscle weakness associated with cardiac conduction blocks, arrhythmias, and restrictive heart failure. It can also lead to myofibrillar destruction with intracytoplasmic accumulation of desmin-reactive deposits in cardiac and skeletal muscle cells. (rEF.: Denise Paulin, D., and Li, Z (2004). Experimental Cell Research 301(1); 1 -7).
Specificity
Immunogen
Application
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Desmin in human heart and human skeletal muscle tissues.
Cell Structure
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected Desmin in 10 µg of human skeletal muscle tissue lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service