Skip to Content
Merck
All Photos(3)

Key Documents

MABT496

Sigma-Aldrich

Anti-PKN2, clone 1D1 Antibody

ascites fluid, clone 1D1, from mouse

Synonym(s):

Serine/threonine-protein kinase N2, Protein kinase C-like 2, Protein-kinase C-related kinase 2, PKN2, PRK2, PRKCL2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$727.00

$727.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$727.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$727.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1D1, monoclonal

species reactivity

mouse, rat, monkey, human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PKN2(5586)

Related Categories

General description

Serine/threonine-protein kinase N2, also known as Protein kinase C-like 2 or Protein-kinase C-related kinase 2, and encoded by the gene name PKN2/PRK2/PRKCL2, is a PKC-related serine/threonine-protein kinase and Rho/Rac effector protein that participates in specific signal transduction responses in the cell. PKN2 has been shown to play an important role in the regulation of cell cycle progression, actin cytoskeleton assembly, cell migration, cell adhesion, tumor cell invasion and transcription activation signaling processes. Protein-kinase-C-related kinases (PRKs) are part of the lipid-regulated protein kinases (PKC) which also include liver PAK & PKN. Human PRK1 and PRK2 share structurally similar catalytic domains, but less similar N-terminal regulatory regions suggesting different regulatory domain functions. PRK1 and PRK2, as well as a third member of this family, PRK3, show distinct patterns of expression in adult tissues. Additionally, the serine-threonine kinase PRK2 can be specifically cleaved by caspase-3 (and/or caspase-3-like subfamily members) during apoptosis. PKN2 is expressed in numerous tumor cell lines, especially in bladder tumor cells.

Immunogen

Purified recombinant fragment of human PKN2 expressed in E. Coli

Application

Anti-PKN2, clone 1D1 Antibody is an antibody against PKN2 for use in western blotting, ELISA, IHC, flow cytometry.
ELISA: A representative lot detected control antigen (100 ng) and antigen (10 ng, 50 ng, 100 ng) in ELISA (Direct).

Immunohistochemistry Analysis: A 1;200-1,000 dilution from a representative lot detected PKN2 in prostate and lung cancer tissues.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected PKN2 in NIH/3T3 cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blotting in PC-12, Cos7, K562, Jurkat, HeLa, A431, C6, NIH/3T3 and HEK293 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected PKN2 in PC-12, Cos7, K562, Jurkat, HeLa, A431, C6, NIH/3T3 and HEK293 cell lysates.

Target description

~140 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hitesh Patel et al.
Scientific reports, 10(1), 3902-3902 (2020-03-05)
PRK1 and PRK2 are two closely related AGC-family serine/threonine protein kinases. Here we demonstrate novel roles for them at cilia and in cancer biology. In both instances serum withdrawal leads to increased activating PRK1 and PRK2 phosphorylation (pPRK1/pPRK2) and their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service