MABT42
Anti-Integrin beta-2 (CD18) Antibody, clone M18/2
clone M18/2, from rat
Synonym(s):
integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), integrin beta chain, beta 2, complement receptor C3 beta-subunit, leukocyte cell adhesion molecule CD18, integrin beta-2, Complement receptor C3 subunit beta, Cell surface adhesion glyco
About This Item
Recommended Products
biological source
rat
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
M18/2, monoclonal
species reactivity
mouse
technique(s)
flow cytometry: suitable
immunoprecipitation (IP): suitable
isotype
IgG2aκ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Itgb2(16414)
General description
Immunogen
Application
Inhibition Analysis: A previous lot of this antibody was shown to inhibit lymphoma metastasis (Zahalka, M. A. et al. (1993) J. Immunol. 150, 4466).
Quality
Flow Cytometry Analysis: 10 µg of this antibody detected Integrin beta-2 in mouse EL4 cells.
Target description
Physical form
Analysis Note
Mouse EL4 cells
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service