Skip to Content
Merck
All Photos(2)

Key Documents

MABT419

Sigma-Aldrich

Anti-CD2AP Antibody, clone 2A2.1

clone 2A2.1, from mouse

Synonym(s):

CD2-associated protein, CD2AP, Adapter protein CMS, Cas ligand with multiple SH3 domains

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$803.00

$803.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$803.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$803.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A2.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD2AP(23607)

General description

CD2-associated protein (UniProt Q9Y5K6; also known as Adapter protein CMS, Cas ligand with multiple SH3 domains) is encoded by the CD2AP (also known as CMS) gene (Gene ID 23607) in human. CD2AP is an adapter protein expressed in all tissues except brain. CD2AP harbors three SH3 domains at its N-terminal, a proline-rich domain, and a C-terminal leucine-zipper domain that mediates CD2AP homodimerization. CD2AP is known to interact with actin-associated proteins, such as cortactin, and actin capping protein CAPZ. The SH3 domains of CD2AP mediate its association to the hypervariable C terminal region of Rac1, allowing CD2AP to act as a linker between Rac1 and the actin-binding proteins cortactin and CAPZ. CD2AP also plays an important role in the formation of the slit diaphragm in the kidney glomerulus between podocyte foot processes by linking the cell-cell adhesion protein nephrin to the cytoskeleton. CD2AP gene mutations are associated with kidney diseases such as sporadic nephrotic syndrome and focal segmental glomerulosclerosis.

Immunogen

GST-tagged recombinant protein corresponding to the C-terminal half of human CD2AP.

Application

Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected CD2AP in human kidney and human colon tissue.
This Anti-CD2AP Antibody, clone 2A2.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of CD2AP.

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected CD2AP in 10 µg of Raji cell lysate.

Target description

~71 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy D Cummins et al.
Journal of cell science, 131(1) (2017-11-28)
Our previous studies of PAWS1 (protein associated with SMAD1; also known as FAM83G) have suggested that this molecule has roles beyond BMP signalling. To investigate these roles, we have used CRISPR/Cas9 to generate PAWS1-knockout U2OS osteosarcoma cells. Here, we show

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service