Skip to Content
Merck
All Photos(1)

Key Documents

MABT416

Sigma-Aldrich

Anti-Nephrin Antibody, clone 3B6.1

clone 3B6.1, from mouse

Synonym(s):

Nephrin, Renal glomerulus-specific cell adhesion receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$563.00

$563.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$563.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$563.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B6.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NPHS1(4868)

General description

Nephrin (UniProt O60500; also known as Renal glomerulus-specific cell adhesion receptor) is encoded by the NPHS1 (also known as NPHN) gene (Gene ID 4868) in human. The transmembrane protein nephrin is an essential component of the podocyte slit diaphragm, the main functional unit of the glomerulus. The renal filtration barrier consists of fenestrated endothelial cells, the glomerular basement membrane (GBM), and the podocytes of epithelial cells. The interdigitating podocyte foot processes are bridged by a 40 nm wide extracellular structure known as the slit diaphragm. Together with the capillary endothelium and GBM, podocytes comprise the glomerular filtration barrier. The GBM functions as a coarse prefilter for large plasma proteins, whereas the slit diaphragm serves as an isoporous ultrafilter with pores smaller than albumin. Nephrin gene mutations are linked to the autosomal recessive disorder congenital nephrotic syndrome of the Finnish type (NPHS1) characterized by massive proteinuria already in utero and nephrosis soon after birth. Both nephrin and the slit diaphragm are reported to be absent in NPHS1 cases. Nephrin is produced with a signal peptide (a.a. 1-22), the removal of which yields the mature protein with a large extracellular region (a.a. 23-1055), followed by a transmembrane segment (a.a. 1056-1076) and a cytoplasmic tail (a.a. 1077-1241). The extracellular region contains eight C-type Ig-like domains (a.a. 27-939), and a Fibronectin type III domain (a.a. 943-1038), while the cytoplasmic tail contains the NPHS2-binding region (a.a. 1160-1241).

Specificity

Clone 3B6.1 recognizes an extracellular epitope in the fifth C2-type Ig-like domain that is present in both spliced isoforms of human nephrin reported by UniProt (O60500).

Immunogen

Epitope: C2-type Ig-like domain 5.
GST-tagged recombinant fragment corresponding to the fifth C2-type Ig-like domain of human Nephrin.

Application

Anti-Nephrin Antibody, clone 3B6.1 is an antibody against Nephrin for use in Western Blotting.
Research Category
Cell Structure
Research Sub Category
Renal Physiology

Quality

Evaluated by Western Blotting in human kidney tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Nephrin in 10 µg of human kidney tissue lysate.

Target description

~185 kDa observed. Target band size appears larger than the calculated molecular weights of 132.5 kDa (isoform 1) and 128.3 kDa (isoform 2; isoform Alpha) due to glycosylation. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service