Recommended Products
antibody form
purified antibody
Quality Level
clone
6/40c
monoclonal
purified by
using protein G
species reactivity
mouse, human
concentration
(Please refer to lot specific datasheet.)
technique(s)
ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG1κ
UniProt accession no.
General description
Recombinant human CEACAM8 extracellular domain.
Specificity
Clone 6/40c specifically detected exogenously expressed human CEACAM8, but not CEACAM1/3/4/5/6/7, on the surface of transfected CHO cells.
Application
Epitope: Extracellular domain
Quality
Evaluated by Western Blotting in PMN cell lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected CEACAM8 in 10 µg of PMN cell lysate.
Western Blotting Analysis: 0.5 µg/mL of this antibody detected CEACAM8 in 10 µg of PMN cell lysate.
Physical properties
~80-140 kDa observed. 35.52/38.15 kDa (mature/pro-form) calculated. The broad and smeary band appearance is consistent with the detection of differentially glycosylated CEACAM8 species.
Physical form
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Storage and Stability
Stable for 1 year at 2-8°C from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service