Skip to Content
Merck
All Photos(1)

Key Documents

MABT1544

Sigma-Aldrich

Anti-Nebulin exon 144 Antibody, clone NEB-9H1

Synonym(s):

NEB;NEM2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$264.00
100 μL
$665.00

$264.00


Estimated to ship on03 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$264.00
100 μL
$665.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$264.00


Estimated to ship on03 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

NEB-9H1, monoclonal

mol wt

calculated mol wt 772.937 kDa
observed mol wt ~47 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NEB(4703)

General description

Nebulin (UniProt: P20929; also known as NEB, NEM2) is encoded by the NEB gene (Gene ID: 4703) in human. Nebulin is a highly conserved, muscle-specific protein that is located in the thin filament of striated muscle. It is involved in maintaining the structural integrity of sarcomeres and the membrane system associated with the myofibrils. It is shown to bind and stabilize F-actin. Nebulin has a highly repetitive protein structure with about 97% of the protein consisting of modules of 30 to 35 amino-acids arranged into simple repeats or super repeats. Each repeat binds to actin along the length of the thin filament. The repeat modules contain conserved SDxxYK consensus sequence that binds F-actin. The N-terminal and the C-terminal ends of Nebulin contain unique protein domains. The C-terminal is anchored in the Z-disc of the muscle sarcomere and contains a conserved Src homology (SH3) domain. Although all known exons in the NEB gene can encode a protein with up to 8,525 amino-acids, alternative splicing may produce somewhat smaller proteins. Most of these alternative splicing events simply remove exons from the nebulin mRNAs, however, all nebulin mRNAs contain either exon 143 or exon 144, but never both. Exon 143 codes for amino acids 5242-5272 and exon 144 codes for amino acids 5277-5307. The sequence encoded by exon 143 shows complete homology between mouse, rat, and human. Studies on NEB knockout mice demonstrate a role for nebulin in establishing lateral connectivity of myofibrils and they lose their tight lateral register on stretch. (Ref.: Lam, L., et al. (2018). Sci. Rep. 8; 15728; Tonino, P., et al. (2010). J. Cell Sci. 123(3); 384-391; Donner, K., et al. (2004). Eur. J. Hum. Genet. 12, 744 751).

Specificity

Clone NEB-9H1 is a mouse monoclonal antibody that detects Nebulin. It targets an epitope with in the sequence encoded by exon 144 (aa 5277-5307).

Immunogen

GST-tagged recombinant fragment corresponding to amino acids encoded by exons 138-144 expressed in E.coli.

Application

Qualtiy Control Testing

Evaluated by Western Blotting in Nebulin recombinant fragments.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected recombinant Nebulin fragments encoded by exons 138-144, but not by exons 138-143.

Tested applications

Immunohistochemistry Applications: A representative lot detected Nebulin exon 144 in Immunohistochemistry applications (Lam, L.T., et al. (2018). Sci Rep. 8(1):15728).

Western Blotting Analysis: A representative lot detected Nebulin exon 144 in Western Blotting applications (Lam, L.T., et al. (2018). Sci Rep. 8(1):15728).

Immunofluorescence Analysis: A representative lot detected Nebulin exon 144 in Immunofluorescence applications (Lam, L.T., et al. (2018). Sci Rep. 8(1):15728).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service