Skip to Content
Merck
All Photos(1)

Key Documents

MABT1505

Sigma-Aldrich

Anti-Plakophilin 1 Antibody, clone 10B2

clone 10B2, from mouse

Synonym(s):

Band 6 protein, B6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$581.00

$581.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$581.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$581.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10B2, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PKP1(5317)

General description

Plakophilin-1 (UniProt: Q13835; also known as Band 6 protein, B6P) is encoded by the PKP1 gene (Gene ID: 5317) in human. Plakophilins are proteins that are involved in linking adherins to cytoskeletal intermediate filaments. They contain numerous armadillo (ARM) repeats and localize to cell desmosomes and nuclei. Plakophilin-1 may be involved in molecular recruitment and stabilization during desmosome formation. It is also shown to be important for tooth development and regulates ZO-1 distribution in the dental epithelium. Two isoforms of Plankophilin-1 have been reported that are produced by alternative splicing. Isoform 1 is expressed in stratified squamous, complex, glandular duct and bladder epithelia and isoform 2 is widely expressed. Plakophilin-1 is shown to be highly expressed in teeth and skin, and was upregulated during tooth development. It is reported to be involved in the regulation of Wnt signaling via its ARM repeat domains in a manner similar to beta-catenin and Wnt activation induces its nuclear translocation. Mutations n PKP1 gene are shown to cause ectodermal dysplasia-skin fragility syndrome that is characterized by cutaneous fragility and congenital ectodermal dysplasia affecting skin, hair and nails. (Ref.: Sprecher, E., et al (2004). J. Invest. Dermatol. 122(3); 647-51; Miyazaki, K et al. (2016). PLoS ONE 11(3): e0152206).

Specificity

Clone 10B2 specifically detects Planophilin-1 and targets an epitope with in the first 27 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
MBP-conjugated recombinant fragment corresponding to the first 235 amino acids from the N-terminal region of human Plakophilin-1.

Application

Anti-Plakophilin 1, clone 10B2, Cat. No. MABT1505, is a mouse monoclonal antibody that detects Plantophilin 1 and has been tested for use in Immunofluorescence and Western Blotting.
Research Category
Cell Structure
Western Blotting Analysis: A representative lot detected Plakophilin 1 in Western Blotting applications (Pigors, M., et. al. (2011). Hum Mol Genet. 20(9):1811-9).

Immunofluorescence Analysis: A representative lot detected Plakophilin 1 in Immunofluorescence applications (Sprecher, E., et. al. (2004). J Invest Dermatol. 122(3):647-51;Pigors, M., et. al. (2011). Hum Mol Genet. 20(9):1811-9).

Quality

Evaluated by Western Blotting in PC3 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Plakophilin 1 in 10 µg of PC3 cell lysate.

Target description

~80 kDa observed; 80.50 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service