Skip to Content
Merck
All Photos(2)

Key Documents

MABT1504

Sigma-Aldrich

Anti-Calponin Antibody, clone CALP

from mouse, clone N3/16, clone CALP

Synonym(s):

Basic calponin, Calponin H1 smooth muscle

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$641.00

$641.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$641.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$641.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CALP, monoclonal
N3/16, monoclonal

species reactivity

human, bovine, porcine

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CNN1(1264)

Related Categories

General description

Calponin-1 (UniProt: P51911; also known as Basic calponin, Calponin H1 smooth muscle) is encoded by the CNN1 gene (Gene ID:1264) in human. Calponin is an actin filament-associated regulatory protein that is expressed in smooth muscle and non-muscle cells of vertebrates. Three isoforms of Calponin have been reported that inhibit actin-activated myosin ATPase activity and stabilize the actin cytoskeleton. Calponin 1 is specifically expressed in smooth muscle cells is involved in fine-tuning of smooth muscle contractility. Calponin-2 is found in multiple cell types, including developing smooth muscles and Calponin-3 is found in the neurons, astrocytes and glial cells. Calponin-1 can bind to actin, calmodulin, troponin C, and tropomyosin. Calponin-1 (and other calponins) has a calponin homology (CH) domain at its N-terminal (aa 28-131) that can function either as a part of actin-binding site or serve as a regulatory structure for actin-binding activity. It also has three calponin-like domains (aa 164-189; 204-229; and 243-268) next to the C-terminal region that are conserved in all calponins. The C-terminal segment varies among different calponins and has differentiated effects on weakening the binding of calponin to F-actin. Deletion of the C-terminal tail segment is shown to enhance the actin-binding and bundling activities of calponins. (Ref.: Liu, R., and Jin, J-P (2016). Gene 585(1); 143-153).

Specificity

Clone CALP specifically detects human Calponin-1.

Immunogen

Crude human uterine muscle extract.

Application

Anti-Calponin, clone CALP , Cat. No. MABT1504, is a mouse monoclonal antibody that detects Calponin-1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected Calponin in human uterus and human prostate tissue sections.
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in human aortic smooth muscle tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Calponin in human aortic smooth muscle tissue lysate.

Target description

~36 kDa observed; 33.17 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Xu et al.
Experimental and therapeutic medicine, 21(4), 322-322 (2021-03-19)
Benign prostatic hyperplasia (BPH) with lower urinary tract symptoms (LUTS) is a common disease among elderly men, for which safe and effective treatment strategies remain limited. The aim of the present study was to explore the potential effects of phosphodiesterase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service