Skip to Content
Merck
All Photos(1)

Key Documents

MABT1335

Sigma-Aldrich

Anti-Tropomyosin 3 Antibody, clone 2G10.2

Synonym(s):

Gamma-tropomyosin, Tropomyosin-3, Tropomyosin-5, TPM3, Tm3

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$266.00
100 μG
$679.00

$266.00


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$266.00
100 μG
$679.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$266.00


Estimated to ship on15 May 2025


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G10.2, monoclonal

mol wt

~30 kDa (Uncharacterized bands may be observed in some lysates.)

purified by

using protein G

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

Gene Information

mouse ... TPM3(117557)

General description

Tropomyosin alpha-3 chain (UniProt: Q63610; also known as Gamma-tropomyosin, Tropomyosin-3, Tropomyosin-5) is encoded by the Tpm3 (also known as Tpm-5, Tpm5) gene (Gene ID: 117557) in rat. The TPM3 gene codes for the slow-twitch skeletal muscle isoform ( s Tm) and at least 9 LMW cytoskeletal isoforms referred to as Tm5NM1 to Tm5NM11. Tropomyosins are dimers of coiled-coil proteins that provide stability to actin filaments and regulate access of other actin-binding proteins. In muscle cells, they regulate muscle contraction by controlling the binding of myosin heads to the actin filament. In non-muscle cells tropomyosins are implicated in stabilizing cytoskeleton actin filaments. Tropomyosins have been implicated in the pathogenesis of cancer where high molecular weight isoforms are consistently down-regulated in transformed cells, while malignant cells display an increased reliance on low molecular weight isoforms. Tropomyosin 3 is a homodimeric protein that can form heterodimers with a beta (TPM2) chain. Its coiled coil structure is formed by 2 polypeptide chains. (Ref.: Glass, JJ et al. (2015). BMC Cancer. 15; 712).

Specificity

Clone 2G10.2 is a mouse monoclonal antibody that specifically detects Tpm3.1 and Tpm3.2. It targets an epitope within 27 amino acids from the C-terminal region.

Immunogen

Diphtheria Toxoid Carrier Protein-conjugated linear peptide corresponding to 27 amino acids from the C-terminal region of rat Tpm3.1 and Tpm3.2

Application

Western Blotting Analysis: 1 µg/mL from a representative lot detected Tropomyosin 3 in human lung tissue lysate.

Immunofluorescence Analysis: A representative lot detected Tropomyosin 3 in Immunofluorescence applications (Schevzov, G., et. al. (2011). Bioarchitecture. 1(4):135-164).

Western Blotting Analysis: A representative lot detected Tropomyosin 3 in Western Blotting applications (Schevzov, G., et. al. (2011). Bioarchitecture. 1(4):135-164).

Quality

Evaluated by Western Blotting in mouse liver tissue lysate. Western Blotting Analysis: 1 µg/mL of this antibody detected Tropomyosin 3 in mouse liver tissue lysate.

Physical form

Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service