Skip to Content
Merck
All Photos(1)

Documents

MABT1324

Sigma-Aldrich

Anti-Keratan sulfate Antibody, clone 373E1

Synonym(s):

KS, Keratosulfate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

373E1, monoclonal

mol wt

observed mol wt ~N/A kDa

purified by

affinity chromatography

species reactivity

chicken, human

species reactivity (predicted by homology)

vertebrates

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgMκ

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Specificity

Clone 373E1 is a mouse monoclonal antibody that detects Keratan sulfate in cartilage tissue.

Immunogen

Purified proteoglycans from embryonic chicken.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Human cartilage tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Keratan Sulfate in human cartilage tissue sections.

Tested Applications

Western Blotting Analysis: A representative lot detected Keratan Sulfate in Western Blotting application (Magro, G., et al. (2003). Am J Pathol. 163(1):183-96).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Keratan Sulfate in Immunoprecipitation application (Magro, G., et al. (2003). Am J Pathol. 163(1):183-96).

Immunohistochemistry Applications: A representative lot detected Keratan Sulfate in Immunohistochemistry application (Magro, G., et al. (2003). Am J Pathol. 163(1):183-96).

ELISA Analysis: A representative lot detected Keratan Sulfate in ELISA application (Magro, G., et al. (2003). Am J Pathol. 163(1):183-96).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Keratan sulfate (also known as keratosulfate) is a widely distributed glycosaminoglycan in tensional and weight-bearing connective tissues, epithelial tissues, and the central and peripheral nervous system. It is attached to a number of proteoglycan and glycoprotein core proteins. Three types of keratan sulfate have been described: KS I, KS II, and KS III depending on their distribution pattern. Keratan sulfate is the only glycosaminoglycan (GAG) that does not contain uronic acid; therefore, it is not cleavable by the eliminative enzymatic cleavage. The biosynthesis of keratan sulfate occurs in two stages. In the first stage, the region binding the core protein with GAG is created followed by the elongation of the chain and its modification. The elongation of keratan sulfate chains of all types occurs by alternate attaching of Gal and GlcNAc residues, catalyzed by β-1,4-galactotransferase and β-1,3-N-acetyloglucosaminotransferase, respectively. Sulfation of keratan sulfate is carried out by at least two sulfotransferases. Gal 6-O-sulfotransferase transfers sulfate to position 6 of Gal residues located in the internal region of keratan sulfate and GlcNAc 6-O-sulfotransferase transfers sulfate to position 6 of GalNAc residues located at the nonreducing terminal of keratan sulfate. Clone 373E1 is a monoclonal antibody that detects a unique sialylation/ conformational-dependent KS epitope that detects a novel glycoprotein complex produced by papillary thyroid carcinoma cells. (Ref.: Magro, G., et al. (2003) Am. J. Pathol. 163(1); 183-196; Caterson, B., et al. (1982). J. Invest. Dermatol. 79 (suppl 1); 45-50; Coleman, PJ., et al. (1998). J. Physiol. 509(3); 695-710).

Physical form

Purified mouse monoclonal antibody IgM in PBS without preservatives.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service