Skip to Content
Merck
All Photos(1)

Key Documents

MABT1323

Sigma-Aldrich

Anti-Cdh1 Antibody, clone AR38.2

clone AR38.2, from mouse

Synonym(s):

Fizzy-related protein homolog, Fzr, Cdh1/Hct1 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$214.00
100 μL
$566.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$214.00
100 μL
$566.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

AR38.2, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CDH1(999)

General description

Fizzy-related protein homolog (UniProt: Q9R1K5; also known as Fzr, Cdh1/Hct1 homolog) is encoded by the Fzr1 (also known as Fyr, Fzr) gene (Gene ID: 56371) in murine species. Cdh1 is a substrate-specific adapter for the anaphase promoting complex/cyclosome (APC/C) E3 ubiquitin-protein ligase complex that associates with the APC/C in late mitosis, in replacement of CDC20, and activates the APC/C during anaphase and telophase. APC/C is activated upon sequential binding to the Cdc20 and Cdh1 adaptor proteins. Cdc20 associates with APC/C during early mitosis and principally regulates mitotic progression, while Cdh1 interacts with the APC/C from late mitosis onwards until the following G1/S transition. The APC/C remains active in degrading substrates to ensure that positive regulators of the cell cycle do not accumulate prematurely. At the G1/S transition Cdh1 is phosphorylated, leading to its dissociation from the APC/C. It has been shown that Cdk-mediated Cdh1 phosphorylation prevents Cdh1 from associating with APC/C prematurely during S/G2 phase, while CDC14B phosphatase-dependent APC/C-Cdh1 activation in response to DNA damage during G2 phase leads to ubiquitination and degradation of Polo-like kinase 1 (Plk1) and prevents entry into mitosis. Following DNA repair, Cdk1 and Plk1 are reactivated and cell cycle progresses from G2 into mitosis phase.

Specificity

Clone AR38.2 detects Cdh1 (Fizzy-related protein homolog) in human and mouse cells.

Immunogen

His-tagged full-length recombinant mouse Fizzy-related protein homolog (Cdh1).

Application

Anti-Cdh1, clone AR38, Cat. No. MABT1323, is a mouse monoclonal antibody that detects Cdh1/FZR1 and has been tested for use in Western Blotting.
Western Blotting Analysis: A representative lot detected Cdh1 in Western Blotting applications (Garzon, J., et. al. (2017). Oncogene. 36(42):5808-5818).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Cdh1 in HeLa cell lysate.

Target description

~54 kDa observed; 54.69 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sahezeel Awadia et al.
The Journal of biological chemistry, 299(6), 104786-104786 (2023-05-06)
The E3 ubiquitin ligase APC/C-Cdh1 maintains the G0/G1 state, and its inactivation is required for cell cycle entry. We reveal a novel role for Fas-associated protein with death domain (FADD) in the cell cycle through its function as an inhibitor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service