Skip to Content
Merck
All Photos(1)

Key Documents

MABT128

Sigma-Aldrich

Anti-JAM-A Antibody, clone BV11

clone BV11, from rat

Synonym(s):

JAM, JAM-1, JAM-A, Ly06, ESTM33, AA638916, 9130004G24, F11r, Junctional adhesion molecule 1, CD321

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$743.00

$743.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BV11, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... F11R(16456)

Related Categories

General description

Junctional adhesion molecule A (JAM-A) is found in endothelial and epithelial tight junctions and is involved in the inflammatory enlistment of mononuclear cells containing LFA-1. JAM-A is linked to atherogenesis, platelet activation, transmigration of leukocytes and angiogenesis. JAM-A is also found to be important in the development of many different organ systems, and is mostly expressed in the epithelial parts and the embryonic vasculature. JAM-A is indicated to play a role in epithelial permeability, inflammation and proliferation in the intestine, aiding in homeostasis.

Immunogen

H5V murine heart capillary endothelial cells.

Application

Detect JAM-A using this Anti-JAM-A Antibody, clone BV11 validated for use in FC, IC & IP.
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in cell extracts from Mouse H5V and PDV cells, and in COS cultures transfected with full length cDNA of mouse JAM-A (Martin-Padura, I., et al. (1998). J Cell Biol. 142(1):117-127).

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in cultured epithelial cells, cremaster muscles, and Schwann Cells (Martin-Padura, I., et al. (1998). J Cell Biol. 142(1):117-127).
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Flow Cytometry in bEnd.3 cells.

Flow Cytometry Analysis: A 1:1,000 dilution of this antibody detected JAM-A in bEnd.3 cells.

Target description

32 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified rat monocolonal IgG2bκ in buffer containing PBS and no preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
bEnd.3 cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catriona T Prendergast et al.
JCI insight, 7(7) (2022-02-23)
Mechanisms governing entry and exit of immune cells into and out of inflamed joints remain poorly understood. We sought herein to identify the key molecular pathways regulating such migration. Using murine models of inflammation in conjunction with mice expressing a
Caio S Bonilha et al.
Immunology letters, 235, 32-40 (2021-05-18)
The junctional adhesion molecule-A (JAM-A) is an adhesion molecule present in the surface of several cell types, such as endothelial cells and leukocytes as well as Dendritic Cells (DC). Given the potential relevance of JAM-A in diverse pathological conditions such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service