Skip to Content
Merck
All Photos(2)

Key Documents

MABS519-AF647

Sigma-Aldrich

Anti-P40 Antibody, clone 11F12.1, Alexa Fluor 647

clone 11F12.1, from mouse, ALEXA FLUOR 647

Synonym(s):

Tumor protein 63, p63, Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, Transformation-related protein 63, TP63, Tumor protein p73-like, p73L, p40, p51, P40

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$689.00

$689.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$689.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$689.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

11F12.1, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TP63(8626)

General description

Tumor protein 63 (UniProt Q9H3D4; also known as Amplified in squamous cell carcinoma, Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p73L, TP63, Transformation-related protein 63, Tumor protein p73-like) is encoded by the TP63 (also known as KET, P63, P73H, P73L, TP73L) gene (Gene ID 8626) in human. Tumor protein 63 is a p53 homologue that can bind p53-responsive elements to upregulate some p53 target genes and share partially overlapping functions with p53. Various functions have been ascribed to TP63 in normal squamous epithelia, including those of the esophagus and TP63 is reported to play an essential role in the maintenance of proliferative potential in epithelial stem cells. TP63 overexpression is found in various squamous epithelia carcinomas and high TP63 expression is associated with an aggressive phenotype and metastasis in many cancers.

Specificity

Expected to react with spliced isoform 2 (DeltaN-alpha, DeltaNp63 alpha, P51delNalpha), 4 (DeltaN-beta, DeltaNp63 beta, P51delNbeta), 6 (DeltaN-gamma, DeltaNp63gamma, P51delNgamma), 8 (DeltaN-delta), 10 (DeltaN-epsilon, DeltaNp73L), 12, but not isoform 1 (TA*-alpha, TAp63alpha, P51B), 3 (TA*-beta, TAp63beta), 5 (TA*-gamma, TAp63gamma, P51A), 7 (TA*-delta, TAp63delta, P51delta), 9 (TA*-epsilon), or 11 (P63 delta).

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human P40 near the N-terminus.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected P40 in HUVEC cells.
The unconjugated version (Cat. No. MABS519) has been shown to work in WB, IH(P).
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Alexa Fluor 647 conjugated Anti-P40 Antibody, clone 11F12.1, is validated for use in Immunocytochemistry (IHC) for the detection of P-40.

Quality

Evaluated by Immunocytochemistry in NIH/3T3 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected P40 in NIH/3T3 cells.

Target description

~40 kDa observed

Physical form

Protein G Purified
Purified mouse conjugate antibody in PBS with 15 mg/ml BSA and 0.1 sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service